Skip to Content
Merck
All Photos(1)

Key Documents

D7692

Sigma-Aldrich

Diethylumbelliferyl phosphate

≥98% (HPLC), oil

Synonym(s):

DEUP, UBP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H17O6P
CAS Number:
Molecular Weight:
312.25
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

oil

color

colorless to yellow

solubility

DMSO: >5 mg/mL

storage temp.

−20°C

SMILES string

CCOP(=O)(OCC)Oc1ccc2C(C)=CC(=O)Oc2c1

InChI

1S/C14H17O6P/c1-4-17-21(16,18-5-2)20-11-6-7-12-10(3)8-14(15)19-13(12)9-11/h6-9H,4-5H2,1-3H3

InChI key

GHGRYKAFUNNHBG-UHFFFAOYSA-N

Application

Diethylumbelliferyl phosphate has been used as a lipase inhibitor to treat wild-type myeloblasts for in vitro neutrophil differentiation assays.

Biochem/physiol Actions

Diethylumbelliferyl phosphate (DEUP), an organophosphate does not block protein kinase activity A in vitro.
Diethylumbelliferyl phosphate is a selective, potent cholesterol esterase inhibitor. It blocks steroidogenesis primarily by preventing cholesterol transport into the mitochondria of steroidogenic cells. Inhibitors of cholesterol esterase are anticipated to limit the absorption of dietary cholesterol. IC50 = 11.6 μM.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Momoko Yamagata et al.
Experimental brain research, 236(5), 1501-1517 (2018-03-23)
We explored the relations between indices of mechanical stability of vertical posture and synergy indices under unexpected perturbations. The main hypotheses predicted higher posture-stabilizing synergy indices and higher mechanical indices of center of pressure stability during perturbations perceived by subjects
Rickson C Mesquita et al.
Biomedical optics express, 1(1), 324-336 (2011-01-25)
Resting state connectivity aims to identify spontaneous cerebral hemodynamic fluctuations that reflect neuronal activity at rest. In this study, we investigated the spatial-temporal correlation of hemoglobin concentration signals over the whole head during the resting state. By choosing a source-detector
Stephen A Brose et al.
Rapid communications in mass spectrometry : RCM, 26(20), 2472-2476 (2012-09-15)
Fatty acids (FA) labeled with (18) O at the carboxyl group, including oxidized species (FA(18) O), are a useful, low-cost, and easy to prepare tool for quantitative and qualitative mass spectrometry (MS) analysis in biological systems. In addition, they are
Jason M Meyer et al.
Journal of lipid research, 53(10), 2081-2091 (2012-07-27)
Macrophage foam cells are a defining pathologic feature of atherosclerotic lesions. Recent studies have demonstrated that at high concentrations associated with hypercholesterolemia, native LDL induces macrophage lipid accumulation. LDL particles are taken up by macrophages as part of bulk fluid
Daniel P Heil et al.
Journal of the International Society of Sports Nutrition, 9, 8-8 (2012-03-22)
Previous research has shown that ingestion of substances that enhance the body's hydrogen ion buffering capacity during high intensity exercise can improve exercise performance. The present study aimed to determine whether the chronic ingestion of an alkalizing supplement, which purports

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service