Skip to Content
Merck
All Photos(1)

Key Documents

L8258

Sigma-Aldrich

Lectin from Wisteria floribunda

lyophilized powder

Synonym(s):

Wisteria floribunda agglutinin, WFA

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.32
Pricing and availability is not currently available.

form

lyophilized powder

potency

<16 μg/mL agglutination activity

composition

Protein, ~95% E1%/280

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

Lectin is a carbohydrate-binding protein found in plant roots, rhizomes, leaves, seeds, and stems.[1][2] Lectin is located in the cytoplasm as well as in the nucleus.[2]

Application

Lectin from Wisteria floribunda has been used:
  • to study the perineuronal nets and parvalbumin nets in mouse brain by immunofluorescence[3]
  • to evaluate the amount of N-acetylgalactosamine (GalNAc) transferred to Gaussia luciferase (GLuc) substrates by microplate assay[4]
  • to study its binding effects on perineuronal nets[5]

Biochem/physiol Actions

Lectin binds with high affinity to the glycans of polysaccharides, glycolipids, and glycoproteins.[1] It is involved in plant defense.[2]
WFA is not blood group specific, but has an affinity for N-acetyl-D-galactosamine.

Packaging

Package size based on protein content

Analysis Note

Agglutination activity is expressed in μg/ml and is determined from serial dilutions in phosphate buffered saline, pH 6.8, of a 1 mg/ml solution. This activity is the lowest concentration to agglutinate a 2% suspension of human blood group A erythrocytes after 1 hr incubation at 25 °C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nausicaä Lannoo et al.
Frontiers in plant science, 5, 397-397 (2014-08-29)
Plants are under constant attack from pathogens and herbivorous insects. To protect and defend themselves, plants evolved a multi-layered surveillance system, known as the innate immune system. Plants sense their encounters upon perception of conserved microbial structures and damage-associated patterns
Lectins, lectin genes, and their role in plant defense.
M J Chrispeels et al.
The Plant cell, 3(1), 1-9 (1991-01-01)
Dorothy Fiete et al.
The Journal of biological chemistry, 287(34), 29194-29203 (2012-06-23)
Two closely related β1,4-N-acetylgalactosaminyltransferases, β4GalNAc-T3 and β4GalNAc-T4, are thought to account for the protein-specific addition of β1,4-linked GalNAc to Asn-linked oligosaccharides on a number of glycoproteins including the glycoprotein hormone luteinizing hormone and carbonic anhydrase-6 (CA6). We have utilized soluble
Markus Morawski et al.
Philosophical transactions of the Royal Society of London. Series B, Biological sciences, 369(1654), 20140046-20140046 (2014-09-17)
Perineuronal nets (PNs) in the brains of tenascin-R-deficient (tn-r(-/-)) mice develop in temporal concordance with those of wild-type (tn-r(+/+)) mice. However, the histological appearance of PNs is abnormal in adult tn-r(-/-) mice. Here, we investigated whether similar defects are also
Baihong Guo et al.
Neurobiology of aging, 96, 223-232 (2020-10-12)
One major pathological process in Alzheimer's disease is mediated by hyperphosphorylated tau, which includes altered microtubules (MTs) and functions associated with tau. A potential way to compensate for altered MT function is to use an MT stabilizer, such as epothilone

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service