Skip to Content
Merck
All Photos(2)

Documents

L5505

Sigma-Aldrich

Lectin from Ulex europaeus (gorse, furze)

lyophilized powder

Synonym(s):

Ulex europaeus agglutinin, UEA-I

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

form

lyophilized powder

Quality Level

potency

<8 μg/mL agglutination activity

composition

Protein, ≥80% biuret

impurities

salt, essentially free

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Ulex europaeus agglutinin has anti-H blood group specificity. There are two types of lectin: UEA I has an affinity for L-fucose and UEA II has an affinity for N,N′-diacetylchitobiose. The mol. wt. of UEA I was initially found to be 170,000 daltons. Later reports indicate that UEA I may form aggregates at neutral and basic pH and that the correct mol. wt. is 68,000 daltons.

Linkage

Conjugates are prepared from affinity purified lectin.

Preparation Note

Purified by affinity chromatography using the method of Pereira.

Analysis Note

Agglutination activity is expressed in μg/ml and is determined from serial dilutions in phosphate buffered saline, pH 6.8, of a 1 mg/ml solution. This activity is the lowest concentration to agglutinate a 2% suspension of human blood group O erythrocytes after 1 hour incubation at 25 °C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D Guy et al.
British journal of cancer, 42(6), 915-921 (1980-12-01)
Present evidence suggests that the cell surface has an important role in metastasis. To examine this idea further, the surface properties of single cells isolated from the primary growths of a liver-metastasizing (ML) and a non-metastasizing (NML) lymphosarcoma were compared
Immunochemical studies on the combining site of the blood group H-specific lectin 1 from Ulex europeus seeds.
M E Pereira et al.
Archives of biochemistry and biophysics, 185(1), 108-115 (1978-01-15)
R T Sawyer et al.
Infection and immunity, 60(3), 1041-1046 (1992-03-01)
The isolated perfused mouse liver model was used to study the effects of various lectins on hepatic trapping and killing of Candida albicans. After mouse livers were washed with 20 to 30 ml of perfusion buffer, 10(6) C. albicans CFU
M Kristensen et al.
Scandinavian journal of medicine & science in sports, 25 Suppl 2, 40-48 (2015-06-18)
We investigated the effect of alpine skiing for 12 weeks on skeletal muscle characteristics and biomarkers of glucose homeostasis and cardiovascular risk factors. Twenty-three patients with a total knee arthroplasty (TKA) were studied 2.9 ± 0.9 years (mean ± SD) after the operation. Fourteen

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service