Skip to Content
Merck
All Photos(1)

Documents

L7900

Sigma-Aldrich

Sodium DL-lactate solution

synthetic, syrup, BioXtra, suitable for mouse embryo cell culture, 60 % (w/w)

Synonym(s):

DL-Lactic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH(OH)COONa
CAS Number:
Molecular Weight:
112.06
Beilstein:
4332999
EC Number:
MDL number:
UNSPSC Code:
12352207
PubChem Substance ID:
NACRES:
NA.75

biological source

synthetic

product line

BioXtra

form

syrup

concentration

60 % (w/w)

technique(s)

cell culture | embryo: suitable

storage temp.

2-8°C

SMILES string

[Na+].CC(O)C([O-])=O

InChI

1S/C3H6O3.Na/c1-2(4)3(5)6;/h2,4H,1H3,(H,5,6);/q;+1/p-1

InChI key

NGSFWBMYFKHRBD-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Lactic acid is an α hydroxyl carboxylic acid produced from pyruvate by the enzyme lactate dehydrogenase (LDH). This mixed isomer lactate has been qualified for use in mouse embryo culture. Lactate regulates pyruvate uptake and metabolism in the preimplantation mouse embryo.

related product

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L J Edwards et al.
Molecular reproduction and development, 50(4), 434-442 (1998-07-21)
Although intracellular pH (pHi), is a regulator of numerous biological processes, it has received relatively little attention with regard to the physiology of the mammalian preimplantation embryo. Interestingly, there is some controversy as to whether the early embryo can recover
M Lane et al.
Biology of reproduction, 62(1), 16-22 (1999-12-28)
This study was an investigation of the interaction of lactate on pyruvate and glucose metabolism in the early mouse embryo. Pyruvate uptake and metabolism by mouse embryos were significantly affected by increasing the lactate concentration in the culture medium. In
Emmi Helle et al.
Frontiers in cell and developmental biology, 9, 715093-715093 (2021-08-24)
Cell-cell interactions are crucial for organ development and function. In the heart, endothelial cells engage in bidirectional communication with cardiomyocytes regulating cardiac development and growth. We aimed to elucidate the organotypic development of cardiac endothelial cells and cardiomyocyte and endothelial
Marek Nalos et al.
Critical care (London, England), 18(2), R48-R48 (2014-03-29)
Acute heart failure (AHF) is characterized by inadequate cardiac output (CO), congestive symptoms, poor peripheral perfusion and end-organ dysfunction. Treatment often includes a combination of diuretics, oxygen, positive pressure ventilation, inotropes and vasodilators or vasopressors. Lactate is a marker of
Wenqian Yuan et al.
Food microbiology, 30(2), 448-452 (2012-03-01)
The aim of the present study was to determine the survival of Salmonella Typhimurium adapted with sodium lactate (NaL), potassium lactate/sodium acetate mixture (KL/NaA) or sodium acetate (NaA) in simulated gastric fluid (SGF) and during heat treatment. NaL-, KL/NaA- and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service