Skip to Content
AD
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Sodium DL-lactate solution
Synonym(s):
DL-Lactic acid sodium salt
Linear Formula:
CH
3
CH(OH)COONa
CAS Number:
72-17-3
Molecular Weight:
112.06
EC Number:
200-772-0
Products
Product Category
buffers (1)
Brand
Sigma-Aldrich (3)
Biological Source
Feature
Formula Weight
Physical Form
Product Line
Markush Class
Markush Group
Technique
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
L4263
Sodium DL-lactate solution,
syrup, BioReagent, 60 % (w/w), synthetic, suitable for cell culture
Expand
View Pricing
L1375
Sodium DL-lactate solution,
syrup, 60 % (w/w), synthetic
Expand
View Pricing
L7900
Sodium DL-lactate solution,
synthetic, syrup, BioXtra, suitable for mouse embryo cell culture, 60 % (w/w)
Expand
View Pricing