Skip to Content
Merck
All Photos(1)

Documents

M5550

Sigma-Aldrich

MEM Amino Acids (50x) solution

Without L-glutamine, liquid, sterile-filtered, BioReagent, suitable for cell culture

Synonym(s):

MEM Amino Acids mixture

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
NACRES:
NA.75

Quality Level

sterility

sterile-filtered

product line

BioReagent

form

liquid

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

Adding supplements of amino acids to media stimulates growth and prolongs the viability of cells in culture. MEM Amino Acids (50x) solution concentrated supplement adds nutrients and reduces the biosynthetic burden on cell in vitro. It has been used in in vitro cultures, co-cultures and incubation media.

Storage and Stability

This product may contain a yellow hue if exposed to light. This is due to the presence of the amino acids tryosine and tryptophan in the solution. The change in color to the liquid solution has no affect to the quality or integrity of the product. For a complete list of components, please see the formulation sheet listed in the "Documents" section of the product page.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Insulin-like growth factor 1 (IGF-1) regulates prolactin, growth hormone, and IGF-1 receptor expression in the pituitary gland of the gilthead sea bream Sparus aurata
Geba KM, et al.
Fish Physiology and Biochemistry, 42(1), 365-377 (2016)
Paleerath Peerapen et al.
Biochimica et biophysica acta, 1864(6), 683-696 (2016-03-16)
Chronic K(+) deficiency can cause hypokalemic nephropathy associated with metabolic alkalosis, polyuria, tubular dilatation, and tubulointerstitial injury. However, effects of acute K(+) deficiency on the kidney remained unclear. This study aimed to explore such effects by evaluating changes in levels
Effect of plasma progesterone on oocyte recovery, oocyte quality,
and early in-vitro developmental competence of embryos in Bos
indicus dairy cows
Saad M, et al.
Animal Reproduction Science, 80-86 (2019)
Ubiquitous marine bacterium inhibits diatom cell division
Van Tol HM, et al.
The ISME Journal, 11(1), 31-42 (2017)
Anne D Thuesen et al.
American journal of physiology. Renal physiology, 307(4), F445-F452 (2014-06-27)
Voltage-gated Ca(2+) (Cav) channels play an essential role in the regulation of renal blood flow and glomerular filtration rate (GFR). Because T-type Cav channels are differentially expressed in pre- and postglomerular vessels, it was hypothesized that they impact renal blood

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service