Skip to Content
Merck
All Photos(4)

Key Documents

L7386

Sigma-Aldrich

Lysostaphin from Staphylococcus staphylolyticus

≥500 units/mg protein, lyophilized powder

Synonym(s):

Glycyl-glycine Endopeptidase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

Product Name

Lysostaphin from Staphylococcus staphylolyticus, lyophilized powder, Protein 50-70 % by biuret, ≥500 units/mg protein

biological source

microbial (Staphylococcus staphylotyticus)

Quality Level

form

lyophilized powder

specific activity

≥500 units/mg protein

mol wt

25 kDa

composition

Protein, 50-70% biuret

packaging

pkg of (sold on basis of mg protein)

solubility

water: 2.00-2.10 mg/mL, clear to slightly hazy, colorless to light yellow

antibiotic activity spectrum

Gram-positive bacteria

application(s)

diagnostic assay manufacturing
diagnostic assay manufacturing

Mode of action

cell wall synthesis | interferes

shipped in

wet ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: peptide

Biochem/physiol Actions

Lysostaphin is a zinc endopeptidase with a molecular weight of approximately 25 kDa. Because lysostaphin cleaves the polyglycine cross-links in the peptidoglycan layer of the cell wall of Staphylococcus species it has been found useful for cell lysis and also as a potential anti-microbial therapeutic.
pH Optimum for activity: ~7.5

Unit Definition

One unit will reduce the turbidity (A620) of a suspension of Staphylococcus aureus cells from 0.250 to 0.125 in 10 min at pH 7.5 at 37°C in a 6.0 ml reaction mixture.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ian R Monk et al.
Nature communications, 10(1), 3067-3067 (2019-07-13)
WalKR (YycFG) is the only essential two-component regulator in the human pathogen Staphylococcus aureus. WalKR regulates peptidoglycan synthesis, but this function alone does not explain its essentiality. Here, to further understand WalKR function, we investigate a suppressor mutant that arose
Judy J J Ou et al.
Frontiers in cellular and infection microbiology, 6, 187-187 (2017-01-14)
Background:Staphylococcus aureus (S. aureus) small colony variants (SCVs) can survive within the host intracellular milieu and are associated with chronic relapsing infections. However, it is unknown whether host invasion rates and immune responses differ between SCVs and their wild-type counterparts.
Catherine Bennett et al.
International forum of allergy & rhinology, 9(8), 918-925 (2019-06-05)
Chronic rhinosinusitis (CRS) is a common disease, often refractory to conventional antimicrobial treatment. In this study we investigate the antimicrobial and anti-inflammatory effects of adding kappa-carrageenan to a commercially available sinus rinse. Kappa-carrageenan was added to Flo CRS and Flo
Dafne Bongiorno et al.
MicrobiologyOpen, 9(5), e1017-e1017 (2020-04-03)
Methicillin-resistant S. aureus has been reported as the main pathogen involved in chronic infections, osteomyelitis, and prosthetic joint infections. The host/pathogen interaction is dynamic and requires several changes to promote bacterial survival. Here, we focused on the internalization and persistence
Igor Belyansky et al.
The Journal of surgical research, 171(2), 409-415 (2011-06-24)
Lysostaphin (LS), a naturally occurring Staphylococcal endopeptidase, has the ability to penetrate biofilm, and has been identified as a potential antimicrobial to prevent mesh infection. The goals of this study were to determine if LS adhered to porcine mesh (PM)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service