Skip to Content
Merck
All Photos(1)

Key Documents

SAE0092

Sigma-Aldrich

Mutanolysin from Streptomyces globisporus ATCC 21553

free of DNA contaminants, suitable for Microbiome research, lyophilized powder, ≥4000 units/mg protein (biuret)

Synonym(s):

Mutanolysin

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.54

biological source

Streptomyces sp. (Streptomyces globisporus ATCC 21553)

Quality Level

form

lyophilized powder

specific activity

≥4000 units/mg protein (biuret)

mol wt

23 kDa
~18.5 kDa

feature

DNA free

concentration

≥40% protein

technique(s)

nucleic acid detection: suitable

suitability

suitable for cell lysis

application(s)

life science and biopharma

shipped in

wet ice

storage temp.

−20°C

Application

Provides gentle cell lysis for the isolation of easily degradable biomolecules and RNA from bacteria. It has been used in the formation of spheroplasts for isolation of DNA.
Purified Mutanolysin SAE0092 undergoes strict quality control testing to ensure the absence of detectable levels of contaminating DNA using 35 cycles PCR amplification of 16S and 18S rDNA using universal primer sets.
Mutanolysin is a muralytic enzyme, suitable for usage in isolation of nucleic acids, from Gram-positive bacteria (e.g. Listeria, Lactobacillus, Lactococcus),(8) and also from other bacteria that are difficult to lyse with lysozyme only.

Biochem/physiol Actions

Mutanolysin cleaves the β-N-acetylmuramyl-(1→4)-N-acetylglucosamine linkage of the bacterial cell wall polymer peptidoglycan-polysaccharide. It lyses Listeria and other gram positive bacteria such as Lactobacillus and Lactococcus.
Mutanolysin is an N-acetylmuramidase. Like lysozyme, it is a muralytic enzyme that cleaves the β-N-acetylmuramyl-(1→4)-N-acetylglucosamine linkage of the bacterial cell wall polymer peptidoglycan-polysaccharide. Its carboxy terminal moieties are involved in the recognition and binding of unique cell wall polymers. Mutanolysin lyses Listeria and other Gram-positive bacteria such as Lactobacillus and Lactococcus.

Features and Benefits

Suitable for difficult-to-lyse samples.

Microbial DNA free, minimizing DNA contamination from reagents in workflow.

Suitable for the isolation of nucleic acids from Gram-positive bacteria, including Listeria, Lactobacillus, and Lactococcus.

Unit Definition

One unit will produce a ΔA600 of 0.01 per minute at pH 6.0 at 37 °C in a 1 mL volume using a suspension of Streptococcus faecalis cell wall as substrate.

Physical form

Lyophilized powder containing Ficoll® and sodium succinate buffer salts

Legal Information

Ficoll is a registered trademark of Cytiva

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G B Calandra et al.
Infection and immunity, 28(3), 1033-1037 (1980-06-01)
Group B streptococci, refractory to previously tested muralysins under physiological conditions, were successfully converted to protoplasts by use of a recently describede N-acetyl muramidase, mutanolysin, derived from a streptomycete. Purified enzyme was effective, but crude preparations, although degrading cell walls
Rebecca M Corrigan et al.
PLoS pathogens, 7(9), e1002217-e1002217 (2011-09-13)
The cell wall is a vital and multi-functional part of bacterial cells. For Staphylococcus aureus, an important human bacterial pathogen, surface proteins and cell wall polymers are essential for adhesion, colonization and during the infection process. One such cell wall
Caroline Meyer Olesen et al.
Microorganisms, 9(7) (2021-08-08)
Investigation of changes in the skin microbiome following treatment of atopic dermatitis (AD) with dupilumab may provide valuable insights into the skin microbiome as a therapeutic target. The aim of this study is to assess changes in the AD skin
Astrid Haaskjold Lossius et al.
Dermatology (Basel, Switzerland), 238(1), 109-120 (2021-04-23)
The pathophysiology in atopic dermatitis (AD) is not fully understood, but immune dysfunction, skin barrier defects, and alterations of the skin microbiota are thought to play important roles. AD skin is frequently colonized with Staphylococcus aureus (S. aureus) and microbial
Martin Bastian Pedersen et al.
Journal of bacteriology, 190(14), 4903-4911 (2008-05-20)
Lactococcus lactis is a widely used food bacterium mainly characterized for its fermentation metabolism. However, this species undergoes a metabolic shift to respiration when heme is added to an aerobic medium. Respiration results in markedly improved biomass and survival compared

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service