Skip to Content
Merck
All Photos(3)

Documents

NA2100

Sigma-Aldrich

GenElute Bacterial Genomic DNA Kits

sufficient for 10 purifications

Synonym(s):

Bacterial Genomic DNA, Gen Elute

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105501
NACRES:
NA.55

usage

sufficient for 10 purifications

storage temp.

15-25°C

General description

The GenElute Bacterial Genomic DNA kit provides a simple and convenient way to isolate pure genomic DNA from gram-negative bacteria. For most gram-positive bacteria, the kit must be used in conjunction with the optional lysozyme (L4919) to effectively lyse the thick peptidoglycan cell walls. A Gram-Positive Lysis Solution is provided as a diluent for preparing the lysozyme stock solutions.

The kit combines the advantages of a silica-based system with a microspin format and eliminates the need for expensive resins, alcohol precipitation, and hazardous organic compounds such as phenol and chloroform.

Application

The purified bacterial genomic DNA is ready for downstream applications such as:
  • restriction endonuclease digestions
  • PCR
  • Southern blots
  • cloning

Features and Benefits

  • Starting material: Up to 1.5 mL of culture
  • Expected yield: Up to 20 μg
  • Elution volume: 400 μl
  • Time required: 70 - 120 min
  • A260/A280 ratio: 1.6 - 1.9
  • No phenol, chloroform, or ethanol precipitation required

Principle

The bacteria are lysed in a chaotropic salt-containing solution to ensure the thorough denaturation of macromolecules. The addition of ethanol causes the DNA to bind when the lysate is spun through a silica membrane into a microcentrifuge tube. After washing to remove the contaminants, the DNA is eluted in 200 μL of a Tris-EDTA solution.

The expected yield of genomic DNA will vary depending on the cell density of the bacterial culture and the bacterial species and strain used. DNA purified with the GenElute kit has an A260/A280 ratio between 1.6 and 1.9 and can be up to 50 kb in length.

Other Notes

For additional information, please see www.sigma-aldrich.com/genomicdna.

Legal Information

GenElute is a trademark of Sigma-Aldrich Co. LLC

Kit Components Also Available Separately

Product No.
Description
SDS

  • W0263Wash Solution 1SDS

  • C2112Column Preparation SolutionSDS

  • P2308Proteinase K from Tritirachium album, lyophilized powder, BioUltra, ≥30 units/mg protein, for molecular biologySDS

  • R6148RNase A solutionSDS

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 3 - Met. Corr. 1 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Storage Class Code

3 - Flammable liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pierre Stallforth et al.
Proceedings of the National Academy of Sciences of the United States of America, 110(36), 14528-14533 (2013-07-31)
Stable multipartite mutualistic associations require that all partners benefit. We show that a single mutational step is sufficient to turn a symbiotic bacterium from an inedible but host-beneficial secondary metabolite producer into a host food source. The bacteria's host is
Ivan M Mukisa et al.
International journal of food microbiology, 160(1), 1-10 (2012-11-13)
Obushera includes four fermented cereal beverages from Uganda namely: Obutoko, Enturire, Ekitiribita and Obuteire, whose microbial diversity has not hitherto been fully investigated. Knowledge of the microbial diversity and dynamics in these products is crucial for understanding their safety and
Susana Delgado et al.
Microbial ecology, 65(3), 763-772 (2013-02-12)
Stomach mucosa biopsies and gastric juices samples of 12 healthy persons were analysed by culturing in selective- and non-selective-rich media. Microbial DNA from four mucosal samples was also amplified by nested PCR using universal bacterial primers, and the 16S rDNA
Christopher S Barker et al.
American journal of orthodontics and dentofacial orthopedics : official publication of the American Association of Orthodontists, its constituent societies, and the American Board of Orthodontics, 143(3), 317-323 (2013-03-05)
Our objective was to determine whether components of fixed orthodontic appliances as received from the manufacturers and after exposure to the clinical environment are free from microbial contamination before clinical use. A pilot molecular microbiologic laboratory study was undertaken at
Cecilia A Silva et al.
Infection and immunity, 80(2), 839-849 (2011-11-16)
Salmonella enterica serovar Enteritidis causes a systemic, typhoid-like infection in newly hatched poultry and mice. In the present study, a library of 54,000 transposon mutants of S. Enteritidis phage type 4 (PT4) strain P125109 was screened for mutants deficient in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service