Skip to Content
Merck
All Photos(2)

Key Documents

M9901

Sigma-Aldrich

Mutanolysin from Streptomyces globisporus ATCC 21553

lyophilized powder, ≥4000 units/mg protein (biuret), Chromatographically purified

Synonym(s):

Mutanolysin

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

biological source

bacterial (Streptomyces globisporus)

Quality Level

form

lyophilized powder

specific activity

≥4000 units/mg protein (biuret)

mol wt

23 kDa

suitability

suitable for cell lysis

application(s)

diagnostic assay manufacturing

shipped in

wet ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

Mutanolysin from Streptomyces globisporus consists of two main lytic enzymes and may be a useful agent for dental caries control.

Application

Mutanolysin has been used in a study to assess lysing and generating protoplasts of dairy streptococci. It has also been used in a study to investigate the conversion of group B streptococci to protoplasts. It provides gentle cell lysis for the isolation of easily degradable biomolecules and RNA from bacteria. It has been used in the formation of spheroplasts for isolation of DNA.
Provides gentle cell lysis for the isolation of easily degradable biomolecules and RNA from bacteria. It has been used in the formation of spheroplasts for isolation of DNA.

Biochem/physiol Actions

Mutanolysin cleaves the β-N-acetylmuramyl-(1→4)-N-acetylglucosamine linkage of the bacterial cell wall polymer peptidoglycan-polysaccharide. It lyses Listeria and other gram positive bacteria such as Lactobacillus and Lactococcus.
Mutanolysin is an N-acetylmuramidase. Like lysozyme, it is a muralytic enzyme that cleaves the β-N-acetylmuramyl-(1→4)-N-acetylglucosamine linkage of the bacterial cell wall polymer peptidoglycan-polysaccharide. Its carboxy terminal moieties are involved in the recognition and binding of unique cell wall polymers. Mutanolysin lyses Listeria and other Gram-positive bacteria such as Lactobacillus and Lactococcus.

Unit Definition

One unit will produce a ΔA600 of 0.01 per minute at pH 6.0 at 37 °C in a 1 mL volume using a suspension of Streptococcus faecalis cell wall as substrate.

Physical form

Lyophilized powder containing Ficoll® and sodium succinate buffer salts

Legal Information

Ficoll is a registered trademark of Cytiva

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Amelia T Soderholm et al.
Frontiers in cellular and infection microbiology, 8, 160-160 (2018-06-06)
Streptococcus pyogenes (Group A Streptococcus; GAS) commonly causes pharyngitis in children and adults, with severe invasive disease and immune sequelae being an infrequent consequence. The ability of GAS to invade the host and establish infection likely involves subversion of host
Pierre Godessart et al.
Nature microbiology, 6(1), 27-33 (2020-11-04)
Gram-negative bacteria are surrounded by a cell envelope that comprises an outer membrane (OM) and an inner membrane that, together, delimit the periplasmic space, which contains the peptidoglycan (PG) sacculus. Covalent anchoring of the OM to the PG is crucial
G B Calandra et al.
Infection and immunity, 28(3), 1033-1037 (1980-06-01)
Group B streptococci, refractory to previously tested muralysins under physiological conditions, were successfully converted to protoplasts by use of a recently describede N-acetyl muramidase, mutanolysin, derived from a streptomycete. Purified enzyme was effective, but crude preparations, although degrading cell walls
Helena Žemličková et al.
Journal of medical microbiology, 67(7), 1003-1011 (2018-06-02)
Purpose. The aim of this study was to characterize serogroup 19 isolates resistant to macrolides and/or penicillin found among pneumococci recovered from cases of invasive and respiratory tract disease in the Czech Republic in 2014.Methods. Pneumococcal isolates of serotypes 19A
Mutanolysin for Improved Lysis and Rapid Protoplast Formation in Dairy Streptococci
Kondo, J. and L. McKay
Journal of Dairy Science, 65, 1428-1431 (1982)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service