Skip to Content
Merck
All Photos(1)

Documents

L6136

Sigma-Aldrich

Lipopolysaccharides from Serratia marcescens

purified by phenol extraction

Synonym(s):

LPS

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352201
NACRES:
NA.25

biological source

Serratia marcescens

Quality Level

form

lyophilized powder

purified by

phenol extraction

impurities

<3% Protein (Lowry)

color

white to yellow cast

solubility

water: 4.90-5.10, faintly hazy to hazy, colorless to faint yellow or tan

shipped in

ambient

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

This product is phenol extracted from Serratia marcescens. The source strain is ATCC 21639.

Biochem/physiol Actions

Lipopolysaccharides (LPS) are localized in the outer layer of the membrane and are, in noncapsulated strains, exposed on the cell surface. They contribute to the integrity of the outer membrane, and protect the cell against the action of bile salts and lipophilic antibiotics.

Preparation Note

The product is soluble in water (5 mg/ml) or cell culture medium (1 mg/ml) yielding a hazy, faint yellow solution. A more concentrated, though still hazy, solution (20 mg/ml) has been achieved in aqueous saline after vortexing and warming to 70-80 oC. Lipopolysaccharides are molecules that form micelles in every solvent. Hazy solutions are observed in water and phosphate buffered saline. Organic solvents do not give clearer solutions. Methanol yields a turbid suspension with floaters, while water yields a homogeneously hazy solution.

Other Notes

To gain a comprehensive understanding of our extensive range of Lipopolysaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mollie R Sorrell et al.
Journal of insect physiology, 126, 104097-104097 (2020-08-14)
Fragile X syndrome (FXS), caused by a mutation in the Fragile X Mental Retardation 1 (FMR1) gene, is a common form of inherited mental retardation. Mutation of the gene leads to a loss of the gene product Fragile X Mental Retardation Protein (FMRP). While a loss
H Heremans et al.
American journal of respiratory and critical care medicine, 161(1), 110-117 (2000-01-05)
Mice given lipopolysaccharide (LPS) intravenously developed lung edema, which was maximum after 6 h. Tumor necrosis factor, interleukin 12 (IL-12), IL-6, and interferon-gamma (IFN-gamma) appeared in the serum, and levels of nitrogen oxide (NO) derivatives were increased in serum and
M G Zenobi et al.
Journal of dairy science, 103(3), 2200-2216 (2020-01-20)
Objectives were to evaluate the effect of prepartum energy intake and peripartal supplementation of ruminally protected choline (RPC) on select indicators of immune status in blood plasma and on lipopolysaccharide-stimulated blood cells ex vivo. At 47 ± 6 d before
Matthew Stephens et al.
Gut microbes, 11(3), 421-432 (2019-06-18)
Patients presenting with Inflammatory bowel disease have been shown to exhibit an altered microbiome in both Crohn's disease and Ulcerative colitis. This shift in the microbial content led us to question whether several of these microbes are important in inflammatory
Victor Meseguer et al.
Nature communications, 5, 3125-3125 (2014-01-22)
Gram-negative bacterial infections are accompanied by inflammation and somatic or visceral pain. These symptoms are generally attributed to sensitization of nociceptors by inflammatory mediators released by immune cells. Nociceptor sensitization during inflammation occurs through activation of the Toll-like receptor 4

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service