Skip to Content
Merck
All Photos(1)

Key Documents

L5543

Sigma-Aldrich

Lipopolysaccharides from Escherichia coli O26:B6

Ready Made solution, 1 mg/mL, 0.2 μm filtered

Synonym(s):

LPS

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352201
NACRES:
NA.25
Pricing and availability is not currently available.

biological source

Escherichia coli (O26:B6)

Quality Level

sterility

0.2 μm filtered

form

aqueous solution

concentration

1 mg/mL

color

colorless to faint yellow

solubility

water: soluble

shipped in

wet ice

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

This product is a 0.2 um filtered aqueous solution containing 1 mg/mL of lipopolysaccharides (LPS) from E. coli serotype O26:B6. This LPS serotype has been used in studies of septic shock.

Application

Lipopolysaccharides (LPSs) are characteristic components of the cell wall of Gram-negative bacteria. LPS and its lipid A moiety stimulate cells of the innate immune system by the Toll-like receptor 4 (TLR4), a member of the Toll-like receptor protein family, which recognizes common pathogen-associated molecular-patterns (PAMPs).

Biochem/physiol Actions

Lipopolysaccharides (LPS) are localized in the outer layer of the membrane and are, in noncapsulated strains, exposed on the cell surface. They contribute to the integrity of the outer membrane, and protect the cell against the action of bile salts and lipophilic antibiotics.

Preparation Note

The LPS used to prepare the solution have been phenol-extracted and chromatographically purified by gel filtration. Lipopolysaccharides prepared by phenol extraction contain up to 60% RNA and less than 1% protein. Subsequent purification by gel filtration chromatography removes much of protein present in the phenol-extracted LPS, but leaves a product that still contains 10-20% nucleic acids. Further purification using ion exchange chromatography yields an LPS product which contains <1% protein and <1% RNA.

Other Notes

To gain a comprehensive understanding of our extensive range of Lipopolysaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Seema Yousuf et al.
Molecular neurobiology, 58(3), 950-963 (2020-10-17)
Vitamin D deficiency (Ddef) alters morphology and outcomes after a stroke. We investigated the interaction of Ddef following post-stroke systemic inflammation and evaluated whether administration of progesterone (P) or vitamin D (D) will improve outcomes. Ddef rats underwent stroke with
Mingwei He et al.
Molecular medicine reports, 21(3), 1163-1171 (2020-01-11)
Approximately 50% of the cases of low back pain (LBP) are attributed to discogenic origin. The causes of discogenic pain are complicated and consist of a complex biochemical cascade. Neovascularization of intervertebral discs (IVDs) is believed to be associated with
Helena Cucak et al.
PloS one, 9(3), e90685-e90685 (2014-03-07)
Type 2 diabetes (T2D) is evolving into a global disease and patients have a systemic low-grade inflammation, yet the role of this inflammation is still not established. One plausible mechanism is enhanced expression and activity of the innate immune system.
Johanna Matilainen et al.
Lipids in health and disease, 19(1), 70-70 (2020-04-15)
Orotic acid (OA) has been intensively utilized to induce fatty liver in rats. Although the capacity of OA to cause steatosis is species-specific, previous in vitro studies indicate that humans could also be susceptible to OA-induced fatty liver. The aim
Dilyana Todorova et al.
EBioMedicine, 62, 103120-103120 (2020-11-27)
With their inherent capability of unlimited self-renewal and unique potential to differentiate into functional cells of the three germ layers, human embryonic stem cells (hESCs) hold great potential in regenerative medicine. A major challenge in the application of hESC-based cell

Questions

  1. What buffer is L5543, LPS in ?

    1 answer
    1. The LPS is in an aqueous solution containing 0.5% triethylamine and pH adjusted to 6.5 using hydrochloric acid.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service