Skip to Content
Merck
All Photos(1)

Key Documents

L5418

Sigma-Aldrich

Lipopolysaccharides from Escherichia coli O55:B5

Ready Made solution, 1 mg/mL

Synonym(s):

LPS

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352201
NACRES:
NA.25

biological source

Escherichia coli (O55:B5)

Quality Level

form

aqueous solution

concentration

1 mg/mL

color

white to faint yellow

solubility

water: 4.90-5.10 mg/mL, faintly hazy to hazy, colorless to faintly yellow

shipped in

wet ice

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

This product is a 0.2 um filtered aqueous solution containing 1 mg/mL of lipopolysaccharides (LPS) from E. coli serotype O55:B5. The source strain is CDC 1644-70. This LPS serotype has been used in studies of septic shock and to induce myocarditis.

Application

Lipopolysaccharides (LPSs) are characteristic components of the cell wall of Gram-negative bacteria. LPS and its lipid A moiety stimulate cells of the innate immune system by the Toll-like receptor 4 (TLR4), a member of the Toll-like receptor protein family, which recognizes common pathogen-associated molecular-patterns (PAMPs).

Biochem/physiol Actions

Lipopolysaccharides (LPS) are localized in the outer layer of the membrane and are, in noncapsulated strains, exposed on the cell surface. They contribute to the integrity of the outer membrane, and protect the cell against the action of bile salts and lipophilic antibiotics.

Preparation Note

The LPS used to prepare the solution have been phenol-extracted and chromatographically purified by gel filtration. Lipopolysaccharides prepared by phenol extraction contain up to 60% RNA and less than 1% protein. Subsequent purification by gel filtration chromatography removes much of protein present in the phenol-extracted LPS, but leaves a product that still contains 10-20% nucleic acids. Further purification using ion exchange chromatography yields an LPS product which contains <1% protein and <1% RNA.

Other Notes

To gain a comprehensive understanding of our extensive range of Lipopolysaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qing Lu et al.
American journal of physiology. Lung cellular and molecular physiology, 301(6), L847-L857 (2011-10-11)
Cigarette smoke (CS) is a major cause of chronic lung and cardiovascular diseases. Recent studies indicate that tobacco use is also a risk factor for acute lung injury (ALI) associated with blunt trauma. Increased endothelial cell (EC) permeability is a
Lauren E Adams et al.
Antioxidants (Basel, Switzerland), 10(3) (2021-04-04)
Therapeutic hypothermia does not improve outcomes in neonatal hypoxia ischemia (HI) complicated by perinatal infection, due to well-described, pre-existing oxidative stress and neuroinflammation that shorten the therapeutic window. For effective neuroprotection post-injury, we must first define and then target CNS
Eric Danilo Pauls Sotelo et al.
Veterinary sciences, 7(2) (2020-05-06)
Several studies in human and equine medicine have produced controversial results regarding the role of dimethylsulfoxide (DMSO) as a therapeutic agent. This study aimed to evaluate the effect of joint lavage with different DMSO concentrations on biomarkers of synovial fluid
Janny C de Grauw et al.
Arthritis research & therapy, 11(2), R35-R35 (2009-03-11)
Inflammation is an important feature of many joint diseases, and levels of cartilage biomarkers measured in synovial fluid may be influenced by local inflammatory status. Little is known about the magnitude and time course of inflammation-induced changes in cartilage tissue
Long-Xiang Su et al.
Chinese medical journal, 132(11), 1336-1343 (2019-03-19)
It has recently been recognized that serum vimentin is elevated in infectious diseases, and that vimentin plays a role in regulating neutrophils and macrophages associated inflammation. However, the mechanisms are unclear. This study was designed to explore the role of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service