Skip to Content
Merck
All Photos(4)

Documents

L5024

Sigma-Aldrich

Lipopolysaccharides from Escherichia coli O127:B8

purified by ion-exchange chromatography, TLR ligand tested

Synonym(s):

LPS

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
MDL number:
UNSPSC Code:
12352201
NACRES:
NA.25

biological source

Escherichia coli (O127:B8)

Quality Level

form

lyophilized powder

purified by

ion-exchange chromatography

impurities

<1% Protein
<1% RNA

color

white to yellow cast

solubility

water: soluble

shipped in

ambient

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

This product is extracted from E. coli serotype O127:B8 and purified by ion exchange. The source strain is ATCC 12740. This LPS serotype has been used in the study of septic shock and to induce NOS in murine macrophages and PAF synthesis in rat glomerular masangial cells.

Application

Lipopolysaccharides (LPSs) are characteristic components of the cell wall of Gram-negative bacteria. LPS and its lipid A moiety stimulate cells of the innate immune system by the Toll-like receptor 4 (TLR4), a member of the Toll-like receptor protein family, which recognizes common pathogen-associated molecular-patterns (PAMPs).

Biochem/physiol Actions

Lipopolysaccharides (LPS) are localized in the outer layer of the membrane and are, in noncapsulated strains, exposed on the cell surface. They contribute to the integrity of the outer membrane, and protect the cell against the action of bile salts and lipophilic antibiotics.

Preparation Note

The product is soluble in water (5 mg/mL) or cell culture medium (1 mg/mL) yielding a hazy, faint yellow solution. A more concentrated, though still hazy, solution (20 mg/mL) has been achieved in aqueous saline after vortexing and warming to 70-80 °C. Lipopolysaccharides are molecules that form micelles in every solvent. Hazy solutions are observed in water and phosphate buffered saline. Organic solvents do not give clearer solutions. Methanol yields a turbid suspension with floaters, while water yields a homogeneously hazy solution.

Other Notes

To gain a comprehensive understanding of our extensive range of Lipopolysaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mingju Cao et al.
Frontiers in cellular neuroscience, 9, 294-294 (2015-08-25)
Neuroinflammation in utero may result in life-long neurological disabilities. The molecular mechanisms whereby microglia contribute to this response remain incompletely understood. Lipopolysaccharide (LPS) or saline were administered intravenously to non-anesthetized chronically instrumented near-term fetal sheep to model fetal inflammation in
Kristen R Hollinger et al.
Journal of Alzheimer's disease : JAD, 77(1), 437-447 (2020-07-18)
Given the emergent aging population, the identification of effective treatments for Alzheimer's disease (AD) is critical. We investigated the therapeutic efficacy of JHU-083, a brain-penetrable glutamine antagonist, in treating AD using the humanized APOE4 knock-in mouse model. Cell culture studies
M Cortes et al.
Scientific reports, 7(1), 10645-10645 (2017-09-08)
Neuroinflammation in utero may result in life-long neurological disabilities. Microglia play a pivotal role, but the mechanisms are poorly understood. No early postnatal treatment strategies exist to enhance neuroprotective potential of microglia. We hypothesized that agonism on α7 nicotinic acetylcholine
Mingju Cao et al.
Frontiers in immunology, 10, 1063-1063 (2019-05-31)
Neuroinflammation in utero may result in lifelong neurological disabilities. Astrocytes play a pivotal role in this process, but the mechanisms are poorly understood. No early postnatal treatment strategies exist to enhance neuroprotective potential of astrocytes. We hypothesized that agonism on
Manman Geng et al.
Clinical immunology (Orlando, Fla.), 220, 108579-108579 (2020-09-01)
Endoplasmic reticulum (ER) stress associated proteins contribute to the pathogenesis of rheumatoid arthritis (RA) through affecting synoviocyte proliferation and proinflammatory cytokine production. The role of DERL3, an ER-associated degradation component, in joint inflammation of RA was explored. Synovial tissues from

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service