Skip to Content
Merck
All Photos(1)

Key Documents

L4513

Sigma-Aldrich

[des-Gly10, D-Ala6]-LH-RH ethylamide acetate salt hydrate

≥97% (HPLC), powder

Synonym(s):

Ethylamide Acetate, Ethylamide Acetate Salt, des-Gly¹⁰ LH-RH

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C56H78N16O12 · xC2H4O2 · yH2O
Molecular Weight:
1167.32 (anhydrous free base basis)
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.32

Assay

≥97% (HPLC)

form

powder

UniProt accession no.

storage temp.

−20°C

SMILES string

O.CC(O)=O.CCNC(=O)[C@@H]1CCCN1C(=O)[C@H](CCCNC(N)=N)NC(=O)[C@H](CC(C)C)NC(=O)[C@@H](C)NC(=O)[C@H](Cc2ccc(O)cc2)NC(=O)[C@H](CO)NC(=O)[C@H](Cc3c[nH]c4ccccc34)NC(=O)[C@H](Cc5c[nH]cn5)NC(=O)[C@@H]6CCC(=O)N6

InChI

1S/C56H78N16O12.C2H4O2.H2O/c1-5-60-54(83)45-13-9-21-72(45)55(84)39(12-8-20-61-56(57)58)66-50(79)40(22-30(2)3)67-47(76)31(4)64-49(78)41(23-32-14-16-35(74)17-15-32)68-53(82)44(28-73)71-51(80)42(24-33-26-62-37-11-7-6-10-36(33)37)69-52(81)43(25-34-27-59-29-63-34)70-48(77)38-18-19-46(75)65-38;1-2(3)4;/h6-7,10-11,14-17,26-27,29-31,38-45,62,73-74H,5,8-9,12-13,18-25,28H2,1-4H3,(H,59,63)(H,60,83)(H,64,78)(H,65,75)(H,66,79)(H,67,76)(H,68,82)(H,69,81)(H,70,77)(H,71,80)(H4,57,58,61);1H3,(H,3,4);1H2/t31-,38+,39+,40+,41+,42+,43+,44+,45+;;/m1../s1

InChI key

YEEHIOYEWOESQR-PFHUABGLSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Amino Acid Sequence

Glp-His-Trp-Ser-Tyr-Ala-Leu-Arg-Pro-NHEt

Application

[des-Gly10, D-Ala6]-LH-RH ethylamide acetate salt hydrate has been used:
  • as an luteinizing hormone-releasing hormone (LHRH) agonist to synchronize a cohort of females rats
  • to synchronize naturally cycling rats
  • to induce ovulation as a gonadotropin-releasing hormone (GnRH) analogue and as a supplement in Galap extender to dilute qualified ejaculate

Biochem/physiol Actions

Potent LH-RH agonist in rats and mice; reversibly delays sexual maturation in rats, stimulates spawning activity in fish. GnRH ( gonadotropin-releasing hormone, Glp-His-Trp-Ser-Tyr-Gly-Leu-Arg-Pro-Gly-NH2), which is also referred as LHRH (luteinizing hormone-releasing hormone) or gonadorelin, is crucial for mammalian reproduction and is released from hypothalamic neurons. It is responsible for the secretion of gonadotropins, luteinizing hormone (LH) and follicle-stimulating hormone (FSH), from the pituitary glands. Mutations in GnRH are associated with normosmic congenital hypogonadotropic hypogonadism.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Amanda B Gillis et al.
PloS one, 16(1), e0245047-e0245047 (2021-01-12)
The aims of this project were to characterize tiger salamander (Ambystoma tigrinum) spermatozoa motility over time, when excreted as either milt or spermic urine prior to packaging into a spermatophore, and to determine the effect of temperature on sperm motility.
E L Mumford et al.
Journal of animal science, 72(1), 174-177 (1994-01-01)
A lack of pituitary LH stores has been implicated as the cause of seasonal anestrus and failure to ovulate during the spring transition period in mares. In this experiment, 40 mares were used to study the effects of GnRH, estrogen
Motility, mitochondrial membrane potential and ATP content of rabbit spermatozoa stored in extender supplemented with GnRH analogue [des-Gly10, D-Ala6]-LH-RH ethylamide
Gogol P, et al.
Polish Journal of Veterinary Sciences, 17(4), 571-575 (2014)
Kari L Chesney et al.
Journal of the American Association for Laboratory Animal Science : JAALAS, 59(3), 282-287 (2020-04-04)
Vaginal cytology is the most common method of monitoring the estrous cycle in rats; however, this test requires specific technical training and can be subject to interpretation. Vaginal impedance offers a quicker and less technically challenging alternative and has been
Ovulation induction in rabbit does by intravaginal administration of the GnRH analogue [des-Gly10, D-Ala6]-LHRH ethylamide: field trial
Reproduction in domestic animals = Zuchthygiene (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service