Skip to Content
Merck
All Photos(1)

Key Documents

CG5

Sigma-Aldrich

Chorionic gonadotropin human

lyophilized powder, vial of ~5,000 IU

Synonym(s):

Choriogonin, HCG

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352209
NACRES:
NA.32
biological source:
human urine
form:
lyophilized powder

biological source

human urine

Quality Level

sterility

sterile-filtered

form

lyophilized powder

mol wt

37.9 kDa

contains

mannitol

packaging

vial of ~5,000 IU

drug control

Home Office Schedule 4.2

impurities

HBsAg, HCV and HIV-1/HIV-2, tested negative

solubility

water: 10 mL/vial, clear to slightly hazy, colorless

UniProt accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

human ... CGA(1081)

Looking for similar products? Visit Product Comparison Guide

General description

Chorionic gonadotropin human (CGA or hCG) is a heterodimer and belongs to the cystine knot growth factor superfamily. It exists as six variants namely hCGn, hCGβ, hCGβn, hCGβcf and hCGα including hCG. CGA is mapped to human chromosome 6q14.3. hCG comprises of α and β chain.
Glycoprotein hormone consisting of a 92-amino acid α-chain which is identical to that of LH, FSH, and TSH; and a distinct 149-amino acid β-chain. Produced by chorionic tissue and responsible for maintaining the corpus luteum during early pregnancy. Also produced by trophoblast cell neoplasms, which are marked by elevated free hCG β-chains and hyperglycosylated hCG.

Application

Chorionic gonadotropin human has been used:
  • to induce oocyte maturation in cultured follicles
  • along with pregnant mare serum gonadotropin (PMSG) to induce superovulation in mice
  • as medium component for fetal testes culture to aid steroidogenic responsiveness

Biochem/physiol Actions

Elevated levels of hCG is observed in trophoblastic cancer. hCG levels are indicators of pregnancy and β subunit is majorly present in urine samples. hCG isoform detection is useful in the diagnosis and management of trophoblastic related diseases including carcinomas.

Reconstitution

When reconstituted with water to a concentration of 1,000 IU/mL, the vial will also contain 0.1 M sodium phosphate buffer (pH approx. 7.2) and 100 mg/mL of mannitol.

Analysis Note

The calculated value is based on the USP method for human Chorionic Gonadotrophin, to meet 5,000 IU per vial, with a tolerance of not less than 80.0% and not more than 125.0% of the potency stated.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Comparison of the oocyte quality derived from two-dimensional follicle culture methods and developmental competence of in vitro grown and matured oocytes
Lee J, et al.
BioMed Research International, 2018 (2018)
Following the course of pre-implantation embryo patterning by non-linear microscopy
Kyvelidou C, et al.
Journal of Structural Biology, 176(3), 379-386 (2011)
Isabelle Mouche et al.
Methods in molecular biology (Clifton, N.J.), 691, 257-269 (2010-10-26)
The Frog Embryo Teratogenesis Assay Xenopus (FETAX) test is a development toxicity screening test. Due to the small amount of compound needed and the capability to study organogenesis in a short period of time (96 h), FETAX test constitutes an
Candidate epitopes for measurement of hCG and related molecules: the second ISOBM TD-7 workshop
Berger P, et al.
Tumor Biology, 34(6), 4033-4057 (2013)
Utility of commonly used commercial human chorionic gonadotropin immunoassays in the diagnosis and management of trophoblastic diseases
Cole LA
Clinical Chemistry, 47(2), 308-315 (2001)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service