Skip to Content
Merck
All Photos(3)

Key Documents

L1881

Sigma-Aldrich

1-Oleoyl-sn-glycero-3-phosphocholine

synthetic, ≥99%

Synonym(s):

(7R,18Z)-4,7-Dihydroxy-N,N,N-trimethyl-10-oxo-3,5,9-trioxa-4-phosphaheptacos-18-en-1-aminium 4-oxide, inner salt, 1-(9Z-Octadecenoyl)-sn-glycero-3-phosphocholine, 1-cis-9-Octadecenoyl-sn-glycero-3-phosphocholine, 1-Oleoyl-2-hydroxy-sn-glycerol-3-phosphocholine, 1-Oleoyl-sn-glycero-3-phosphorylcholine, 3-sn-Lysophosphatidylcholine, 1-oleoyl, L-γ-Oleoyl-α-lysolecithin, Lysolecithin, oleoyl, PC(18:1(9Z)/0:0), PC(18:1/0:0)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H52NO7P
CAS Number:
Molecular Weight:
521.67
Beilstein:
4033747
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.25

biological source

synthetic

Assay

≥99%

form

powder

functional group

phospholipid

lipid type

phosphoglycerides

shipped in

ambient

storage temp.

−20°C

SMILES string

O[C@](COP([O-])(OCC[N+](C)(C)C)=O)([H])COC(CCCCCCC/C=C\CCCCCCCC)=O

InChI

1S/C26H52NO7P/c1-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-26(29)32-23-25(28)24-34-35(30,31)33-22-21-27(2,3)4/h12-13,25,28H,5-11,14-24H2,1-4H3/b13-12-/t25-/m1/s1

InChI key

YAMUFBLWGFFICM-PTGWMXDISA-N

Looking for similar products? Visit Product Comparison Guide

Application

1-Oleoyl-sn-glycero-3-phosphocholine may be used as a reference for the analysis of lysolecithins extracted from tissues and cell membranes.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B M Ross et al.
Journal of neurochemistry, 63(5), 1839-1848 (1994-11-01)
Lysophospholipids are generated during the turnover and breakdown of membrane phospholipids. We have identified and partially characterized three enzymes involved in the metabolism of lysophospholipids in human brain, namely, lysophospholipase, lysophospholipid:acyl-CoA acyltransferase (acyltransferase), and lysophospholipid:lysophospholipid transacylase (transacylase). Each enzyme displayed
Gerd Schmitz et al.
Atherosclerosis, 208(1), 10-18 (2009-07-03)
Lysophosphatidylcholine (LPC) is a major plasma lipid that has been recognized as an important cell signalling molecule produced under physiological conditions by the action of phospholipase A(2) on phosphatidylcholine. LPC transports glycerophospholipid components such as fatty acids, phosphatidylglycerol and choline
Lynda M Foulds et al.
Biology of reproduction, 79(3), 525-536 (2008-05-30)
The ability of the gametes to escape detection by the immune system is vital to successful human reproduction. Furthermore, the observed capacity of the testis in some species to support tissue grafts without rejection (immunological privilege) indicates that spermatogenic cells
Swati Anand et al.
Biomarkers in medicine, 9(6), 563-575 (2015-06-17)
We sought serum biomarkers predictive of pre-eclampsia (PE). Sera obtained at 12-14 weeks of pregnancy from 24 cases who later developed PE and 24 controls with uncomplicated pregnancies were processed and analyzed using a serum proteomic approach. Many statistically significant
Yoshinori Okamoto et al.
Toxicology letters, 288, 65-70 (2018-02-20)
Estrogen is reported to be involved in mammary tumorigenesis. To unveil metabolic signatures for estrogen-induced mammary tumorigenesis, we carried out serum metabolomic analysis in an estrogen-induced mammary tumor model, female August Copenhagen-Irish/Segaloff (ACI/Seg) rats, using liquid chromatography-mass spectrometry. In contrast

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service