Skip to Content
Merck
All Photos(2)

Documents

O0383

Sigma-Aldrich

N-Oleoylethanolamine

~98% (TLC)

Synonym(s):

N-(Hydroxyethyl)oleamide, N-(cis-9-Octadecenoyl)ethanolamine, OEA, oleoylethanolamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H39NO2
CAS Number:
Molecular Weight:
325.53
EC Number:
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.83

Assay

~98% (TLC)

form

powder

solubility

chloroform: 20 mg/mL, clear, colorless

storage temp.

−20°C

SMILES string

O=C(CCCCCCC/C=C\CCCCCCCC)NCCO

InChI

1S/C20H39NO2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-20(23)21-18-19-22/h9-10,22H,2-8,11-19H2,1H3,(H,21,23)/b10-9-

InChI key

BOWVQLFMWHZBEF-KTKRTIGZSA-N

Gene Information

General description

Oleoylethanolamide (OEA) is an endogenous fatty acid ethanolamine. Small intestine cells, adipose tissues, neurons and astrocytes produces OEA.

Application

N-Oleoylethanolamine has been used to study its effects on glucagon-like peptide (GLP)-1RA-mediated anorectic signaling and weight loss.

Biochem/physiol Actions

Oleoylethanolamide (OEA) is a lipid mediator, which helps to control various biological functions, like food intake. It modulates the gene expression in the small intestine. OEA has the ability to initiate several receptors, such as cannabinoid receptor type 1 (CB1), peroxisome proliferator-activated receptors (PPARs) and G protein-coupled receptor 119 (GPR119).
Peroxisome proliferator-activator receptor α (PPARα) agonist

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Gastrointestinal Tract and Control of Food Intake
Physiology of the Gastrointestinal Tract, 2, 1963-1978 (2012)
Oleoylethanolamide modulates glucagon-like peptide-1 receptor agonist signaling and enhances exendin-4-mediated weight loss in obese mice
Brown JD, et al.
American Journal of Physiology. Regulatory, Integrative and Comparative Physiology, 315(4), R595-R608 (2018)
Byung-Koo Son et al.
Nanomaterials (Basel, Switzerland), 9(7) (2019-07-13)
A mixture of nanothin exfoliated (NTE) graphite and urea (CO(NH2)2) powder was treated with radio frequency (RF) thermal plasma to achieve in situ purification and nitrogen doping of NTE graphite using the high-temperature flame of the RF plasma. Reactive species
Xiaotong Diao et al.
Nature communications, 13(1), 2529-2529 (2022-05-10)
Hypoxia-inducible factors (HIFs) are α/β heterodimeric transcription factors modulating cellular responses to the low oxygen condition. Among three HIF-α isoforms, HIF-3α is the least studied to date. Here we show that oleoylethanolamide (OEA), a physiological lipid known to regulate food
Mehdi Boutagouga Boudjadja et al.
Current biology : CB, 32(21), 4699-4706 (2022-10-02)
Loss of appetite and negative energy balance are common features of endotoxemia in all animals and are thought to have protective roles by reducing nutrient availability to host and pathogen metabolism. Accordingly, fasting and caloric restriction have well-established anti-inflammatory properties.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service