Skip to Content
Merck
All Photos(1)

Documents

C9253

Sigma-Aldrich

Cholesteryl oleate

≥98% (HPLC; detection at 205 nm)

Synonym(s):

3β-Hydroxy-5-cholestene 3-oleate, 5-Cholesten-3β-ol 3-oleate, Cholesteryl cis-9-octadecenoate, Oleic acid cholesteryl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C45H78O2
CAS Number:
Molecular Weight:
651.10
Beilstein:
2343071
EC Number:
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.77

biological source

synthetic (organic)

Assay

≥98% (HPLC; detection at 205 nm)

form

powder

mp

44-47 °C (lit.)

functional group

ester

shipped in

ambient

storage temp.

−20°C

SMILES string

[H]\C(CCCCCCCC)=C(/[H])CCCCCCCC(=O)O[C@H]1CC[C@@]2(C)C(C1)=CC[C@]3([H])[C@]2([H])CC[C@]4(C)[C@]([H])(CC[C@@]34[H])[C@H](C)CCCC(C)C

InChI

1S/C45H78O2/c1-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-25-43(46)47-38-30-32-44(5)37(34-38)26-27-39-41-29-28-40(36(4)24-22-23-35(2)3)45(41,6)33-31-42(39)44/h14-15,26,35-36,38-42H,7-13,16-25,27-34H2,1-6H3/b15-14-/t36-,38+,39+,40-,41+,42+,44+,45-/m1/s1

InChI key

RJECHNNFRHZQKU-RMUVNZEASA-N

Looking for similar products? Visit Product Comparison Guide

Application

Cholesteryl oleate was used to study the ability of silicone hydrogel contact lenses to absorb tear lipids, cholesterol and cholesterol esters. It was also used in the development of amperometric cholesterol biosensors.

Biochem/physiol Actions

Cholesterol in plasma is present in an esterified form such as Cholesteryl oleate. The liver is a primary source of cholesteryl oleate that enriches the LDL particles when released into plasma. The secretion of lipoproteins enriched with cholesteryl oleate has close correlation with the pathogenesis of coronary artery atherosclerosis.

Preparation Note

Cholesteryl oleate yields a clear, colorless solution in chloroform at 100 mg/ml.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J A Hamilton et al.
The Journal of biological chemistry, 258(21), 12821-12826 (1983-11-10)
The incorporation of cholesteryl oleate and triolein into phospholipid vesicles was studied in cosonicated mixtures of 94 weight % egg phosphatidylcholine and 6 weight % neutral lipid (0-6% triolein and 6-0% cholesteryl oleate). 13C NMR spectroscopy was used to quantitate
L L Rudel et al.
The Journal of clinical investigation, 100(1), 74-83 (1997-07-01)
Relationships among plasma lipoprotein cholesterol, cholesterol secretion by the isolated, perfused liver, and coronary artery atherosclerosis were examined in African green monkeys fed diets containing cholesterol and 35% of calories as fat enriched in polyunsaturated, monounsaturated, or saturated fatty acids.
Marta Overchuk et al.
ACS nano, 13(4), 4560-4571 (2019-03-28)
Lipoprotein mimetic nanostructures, which consist of an amphiphilic lipid shell, a hydrophobic core, and an apolipoprotein mimetic peptide, serve as a versatile platform for the design of drug delivery vehicles as well as the investigation of supramolecular assemblies. Porphyrin incorporation
Design of an Interference-Free Cholesterol Amperometric Biosensor Based on the Electrosynthesis of Polymeric Films of Diaminonaphthalene Isomers
Garcia-Ruiz E et al
Electroanalysis, 16, 497-504 (2004)
Chendong Yang et al.
The Journal of clinical investigation, 114(6), 813-822 (2004-09-17)
The ABC transporters ABCG5 and ABCG8 limit absorption and promote excretion of dietary plant sterols. It is not known why plant sterols are so assiduously excluded from the body. Here we show that accumulation of plant sterols in mice lacking

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service