Skip to Content
Merck
All Photos(1)

Key Documents

I3766

Sigma-Aldrich

Isoliquiritigenin

powder

Synonym(s):

(E)-1-(2,4-Dihydroxyphenyl)-3-(4-hydroxyphenyl)-2-propen-1-one, 4,2′,4′-Trihydroxychalcone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H12O4
CAS Number:
Molecular Weight:
256.25
Beilstein:
1914296
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.77

form

powder

Quality Level

color

yellow to orange

solubility

H2O: <0.1 mg/mL
methanol: 10 mg/mL
DMSO: 20 mg/mL

storage temp.

2-8°C

SMILES string

Oc1ccc(cc1)\C=C\C(=O)c2ccc(O)cc2O

InChI

1S/C15H12O4/c16-11-4-1-10(2-5-11)3-8-14(18)13-7-6-12(17)9-15(13)19/h1-9,16-17,19H/b8-3+

InChI key

DXDRHHKMWQZJHT-FPYGCLRLSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

Isoliquiritigenin is an aromatic ketone and belongs to the chalcone group of compound. It is derived from licorice and is a component in medicine and food.

Application

Isoliquiritigenin has been used:
  • as a bone morphogenetic protein (BMP) agonist in zebrafish embryos
  • as a guanylyl cyclase activator in zebrafish embryos
  • to test its antitumor and antiviral effects against Epstein-Barr virus-associated gastric carcinoma

Biochem/physiol Actions

Isoliquiritigenin is a soluble guanylyl cyclase activator and possesses antitumor activity. It also possesses antioxidant, antiplatelet and estrogenic properties.

Features and Benefits

This compound is a featured product for Cyclic Nucleotide research. Click here to discover more featured Cyclic Nucleotide products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Caution

Photosensitive, hygroscopic

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wdr68 mediates dorsal and ventral patterning events for craniofacial development
Alvarado E, et al.
PLoS ONE, 11(11), e0166984-e0166984 (2016)
Lei Yang et al.
Food chemistry, 138(1), 173-179 (2012-12-26)
An ionic liquid-based ultrasonic-assisted extraction (ILUAE) method had been used for the effective extraction of isoliquiritigenin (IQ), liquiritin (LQ) and glycyrrhizic acid (GA) from licorice. The ionic liquids with different cations and anions were investigated in this work and 0.5
Xuan Yuan et al.
Recent patents on anti-cancer drug discovery, 8(2), 191-199 (2012-09-12)
Isoliquiritigenin (ISL), a licorice chalconoid, is a bioactive agent with chemopreventive potential that has been patented for tumor treatment in China. This study investigated the mechanisms of ISL-induced apoptosis in ovarian carcinoma SKOV-3 cells. Cell viability was evaluated using a
S Yamamoto et al.
Carcinogenesis, 12(2), 317-323 (1991-02-01)
A topical application of a chalcone derivative, 4,2',4'-trihydroxychalcone (isoliquiritigenin) inhibited epidermal ornithine decarboxylase (ODC) induction and ear edema formation, i.e. inflammation, caused by a topical application of 12-O-tetradecanoylphorbol-13-acetate (TPA) in CD-1 mice. In addition, isoliquiritigenin potently inhibited 7,12-dimethylbenz[alpha]anthracene (DMBA)-initiated and
Kai-Lee Wang et al.
Expert opinion on therapeutic targets, 17(4), 337-349 (2013-01-19)
Isoliquiritigenin (ISL) is a natural phenolic compound extracted from licorice. Previous studies have shown that ISL is a potent antioxidant with anti-inflammatory and antitumor activities. The anti-invasive activity of ISL was still unclear. The actual causes of death for most

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service