Skip to Content
Merck
All Photos(4)

Documents

G6649

Sigma-Aldrich

Genistein

≥98% (HPLC), powder, tyrosine protein kinase inhibitor

Synonym(s):

4′,5,7-Trihydroxyisoflavone, 5,7-Dihydroxy-3-(4-hydroxyphenyl)-4H-1-benzopyran-4-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H10O5
CAS Number:
Molecular Weight:
270.24
Beilstein:
263823
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

product name

Genistein, synthetic, ≥98% (HPLC), powder

biological source

synthetic

Assay

≥98% (HPLC)

form

powder

color

off-white to yellow

mp

297.0-298.0 °C

solubility

DMSO: soluble
ethanol: soluble

storage temp.

−20°C

SMILES string

Oc1ccc(cc1)C2=COc3cc(O)cc(O)c3C2=O

InChI

1S/C15H10O5/c16-9-3-1-8(2-4-9)11-7-20-13-6-10(17)5-12(18)14(13)15(11)19/h1-7,16-18H

InChI key

TZBJGXHYKVUXJN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Genistein is an isoflavonoid present among Leguminosae family of plants. It is structurally similar to estrogen (estradiol-17β).

Application

Genistein has been used:
  • as a test compound to access its estrogeniic acivity
  • as an oestrogenic ligand to carry out identical reporter gene activation assays and also used to examine the binding of genistein to hERβ
  • as an endocytosis inhibitor to test the possible effect of endocytosed advanced glycation end - bovine serum albumin (AGE-BSA) on lysosomes

Biochem/physiol Actions

Genistein is essential for plant-microbe interaction. It acts as a natural chemoprotectant and helps in preventing tumor progression and heart disorders. Genistein functionally shares estrogen′s action. It is useful in evaluating nutraceuticals and functional foods. Genistein exhibits antimicrobial action and can serve as a phytoalexin and phytoanticipin.
Inhibitor of tyrosine protein kinase; competitive inhibitor of ATP in other protein kinase reactions. Antiangiogenic agent, down-regulates the transcription of genes involved in controlling angiogenesis.

Features and Benefits

This compound is a featured product for Kinase Phosphatase Biology research. Click here to discover more featured Kinase Phosphatase Biology products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound is featured on the Chloride Channels and Met pages of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Genistein
Dixon RA and Ferreira D
Phytochemistry, 60(3), 205-211 (2002)
Autophagy-lysosome pathway in renal tubular epithelial cells is disrupted by advanced glycation end products in diabetic nephropathy
Liu WJ, et al.
Test, jbc-M115 (2015)
Cloning of oestrogen receptor beta from Old and New World primates: identification of splice variants and functional analysis
Sierens JE, et al.
Journal of Molecular Endocrinology, 32(3), 703-718 (2004)
Assaying estrogenicity by quantitating the expression levels of endogenous estrogen-regulated genes.
Jorgensen M, et al.
Environmental Health Perspectives, 108(5), 403-403 (2000)
Haolong Li et al.
Oncotarget, 6(24), 20474-20484 (2015-05-27)
Although the new generation of androgen receptor (AR) antagonists like enzalutamide (ENZ) prolong survival of metastatic castration-resistant prostate cancer (CRPC), AR-driven tumors eventually recur indicating that additional therapies are required to fully block AR function. Since DNA topoisomerase II (Topo

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service