Skip to Content
Merck
All Photos(1)

Key Documents

G9548

Sigma-Aldrich

Glabridin

≥98% (HPLC), powder, isoflavan

Synonym(s):

4-[(3R)-3,4-Dihydro-8,8-dimethyl-2H,8H-benzo[1,2-b:3,4-b′]dipyran-3-yl]-1,3-benzenediol, Glabridin, (R)-4-(3,4-Dihydro-8,8-dimethyl-2H,8H-benzo[1,2-b:3,4-b′]dipyran-3-yl)-1,3-benzenediol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H20O4
CAS Number:
Molecular Weight:
324.37
Beilstein:
7141956
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77
Pricing and availability is not currently available.

Product Name

Glabridin, ≥98% (HPLC), powder

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to light brown

solubility

DMSO: 5 mg/mL, clear (warmed)

storage temp.

room temp

SMILES string

CC1(C)Oc2ccc3C[C@@H](COc3c2C=C1)c4ccc(O)cc4O

InChI

1S/C20H20O4/c1-20(2)8-7-16-18(24-20)6-3-12-9-13(11-23-19(12)16)15-5-4-14(21)10-17(15)22/h3-8,10,13,21-22H,9,11H2,1-2H3/t13-/m0/s1

InChI key

LBQIJVLKGVZRIW-ZDUSSCGKSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Glabridin has been used:
  • to analyze its transthyretin (TTR)-stabilizing effect in plasma of healthy volunteers[1]
  • as a standard in proton nuclear magnetic resonance (1H NMR) to study the interaction between glabridin and human serum albumin using multi-spectroscopic and molecular docking techniques[2]
  • to study its effect on nucleotide-binding oligomerization domain-like receptor pyrin-domain-containing protein 3 (NLRP3) inflammasome activation[3]

Biochem/physiol Actions

Glabridin is a bio-available isoflavan isolated from licorice (Glycyrrhiza glabra L.) root extract. Glabridin has been reported to possess various biological activities including strong antioxidant activity, antioxidant against LDL oxidation, anti-Helicobacter pylori properties, estrogen-like activity and antinephritic and radical scavenging activities. Also it appears to inhibit serotonin re-uptake, melanogenesis, inflammation and cytochrome P450 3A4, 2B6, and 2C9.
Glabridin is a main component of glavonoid, also known as licorice-derived flavonoid oil, which is a natural supplement used in traditional medicine and herbal medicine.[1]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A novel fluorescent recombinant cell-based biosensor for screening NLRP3 inflammasome inhibitors
Ye Y, et al.
Sensors and Actuators B, Chemical, 301, 126864-126864 (2019)
Fumiki Aoki et al.
Journal of the American College of Nutrition, 26(3), 209-218 (2007-07-20)
Licorice flavonoids have various physiological activities such as abdominal fat-lowering, hypoglycemic and antioxidant effects. Licorice flavonoid oil (LFO: Kaneka Glavonoid Rich Oil) is a new dietary ingredient containing licorice flavonoids dissolved in medium-chain triglycerides (MCT). Glabridin is one of the
Hiroaki Matsushita et al.
Heliyon, 7(10), e08101-e08101 (2021-10-26)
Transthyretin (TTR) is an amyloidogenic protein associated with hereditary and nonhereditary transthyretin amyloidoses (ATTR). Dissociation of the tetramer of TTR to the monomer induces TTR misfolding, which leads to amyloid fibril formation and triggers the onset of ATTR amyloidosis. Stabilizers
Structural insights into the binding behavior of isoflavonoid glabridin with human serum albumin
Razzak MA, et al.
Food Hydrocolloids, 91, 290-300 (2019)
Itamar Yehuda et al.
Phytotherapy research : PTR, 25(5), 659-667 (2010-11-06)
The risk of death from cardiovascular diseases (CVDs), which are exacerbated by oxidative stress, is higher in diabetic women. This phenomenon has been attributed to the loss of estradiol-vascular protection. Such knowledge led us to examine the potential of glabridin

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service