Skip to Content
Merck
All Photos(1)

Key Documents

P0020

Sigma-Aldrich

Pristimerin

Synonym(s):

(9b,13a,14b,20a)-3-Hydroxy-9,13-dimethyl-2-oxo-24,25,26 -trinoroleana-1(10),3,5,7-tertraen-29-oic acid methyl ester, Celastrol methyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H40O4
CAS Number:
Molecular Weight:
464.64
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

Quality Level

form

powder

storage condition

protect from light

color

orange

solubility

DMSO: ≥5 mg/mL

storage temp.

−20°C

SMILES string

COC(=O)[C@]1(C)CC[C@]2(C)CC[C@]3(C)C4=CC=C5C(C)=C(O)C(=O)C=C5[C@]4(C)CC[C@@]3(C)C2C1

InChI

1S/C30H40O4/c1-18-19-8-9-22-28(4,20(19)16-21(31)24(18)32)13-15-30(6)23-17-27(3,25(33)34-7)11-10-26(23,2)12-14-29(22,30)5/h8-9,16,23,32H,10-15,17H2,1-7H3/t23-,26-,27-,28+,29-,30+/m1/s1

InChI key

JFACETXYABVHFD-WXPPGMDDSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Pristimerin has been used as an anti-tumor agent to study its effects on conditionally reprogrammed patient-derived lung adenocarcinoma cells (CRLCs). It has also been used as an anti-tumor agent to study its effects on conditionally reprogrammed patient derived-primary hepatocellular carcinoma cells (CRHCs).

Biochem/physiol Actions

More active than euphol against MGL, better activity with rat neurons, but less selective relative to similar enzymes.
More active than euphol against MGL, better activity with rat neurons, but less selective relative to similar enzymes. First MGL inhibitor to act reversibly, several others covalently bind to cysteine residues. Other studies involve multiply mylome, pristimerin inhibits NF-κB activation via inhibition of IKK-α or IKK-β. It is the methyl ester of celastrol (C0869).
Pristimerin is a quinone methide triterpenoid found abundantly in Celastraceae and Hippocrateaceae families. It shows anti-inflammatory, anti-malarial, anti-bacterial, and insecticidal properties. Pristimerin exhibits anti-cancer and anti-proliferative activity by affecting vasculogenesis, apoptosis, autophagy, migration, and invasion of tumor cells. It has been studied to exhibit therapeutic effects against leukemia, glioma, breast cancer, prostate cancer, oral cancer, and lung cancer.

Features and Benefits

This compound is featured on the Cannabinoid Receptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class Code

11 - Combustible Solids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Du-Qiang Luo et al.
Pest management science, 61(1), 85-90 (2004-12-14)
Pristimerin and celastrol isolated from the roots of Celastrus hypoleucus (Oliv) Warb f argutior Loes exhibited inhibitory effects against diverse phytopathogenic fungi. Pristimerin and celastrol were found to inhibit the mycelial growth of Rhizoctonia solani Kuhn and Glomerella cingulata (Stonem)
Xianmin Mu et al.
Molecules (Basel, Switzerland), 17(6), 6854-6868 (2012-06-07)
Pristimerin is a triterpenoid isolated from Celastrus and Maytenus spp. that has been shown to possess a variety of biological activities, including anti-cancer activity. However, little is known about pristimerin's effects on tumor angiogenesis. In this study, we examined the
Joo-Yun Byun et al.
Molecular pharmacology, 76(4), 734-744 (2009-07-04)
Naturally occurring triterpenoid compounds have long been used as anti-inflammatory, antimalarial, and insecticidal agents. It has become evident that some of the natural or synthetic triterpenoids have promising clinical potential as both a therapeutic and chemopreventive agent for cancer. However
V M Dirsch et al.
European journal of pharmacology, 336(2-3), 211-217 (1998-01-24)
Inducible nitric oxide synthase dependent production of nitric oxide (NO) plays an important role in inflammation. We investigated whether pristimerin ((20alpha)-3-hydroxy-2-oxo-24-nor-friedela-1(10),3,5,7-te traen-carboxylic acid-(29)-methylester), an antitumoral, antimicrobial as well as anti-inflammatory plant compound, has an effect on the inducible NO synthase
Da-Young Eum et al.
Anti-cancer drugs, 22(8), 763-773 (2011-06-07)
A combined treatment with conventional chemotherapies can enhance the effectiveness of chemotherapeutic agents against cancers. Here, we have shown that the naturally occurring triterpenoids synergistically enhance the response of cervical cancer cells to taxol. Of the triterpenoid compounds, pristimerin enhanced

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service