Skip to Content
Merck
All Photos(1)

Documents

H6909

Sigma-Aldrich

Hydrocortisone solution

50 μM, sterile-filtered, BioXtra, suitable for cell culture

Synonym(s):

17-Hydroxycorticosterone, Cortisol

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.77

biological source

non-animal source

sterility

sterile-filtered

product line

BioXtra

form

liquid

concentration

50 μM

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

pH

5.0- 7.0

shipped in

dry ice

storage temp.

−20°C

SMILES string

C[C@]12CCC(=O)C=C1CC[C@H]3[C@@H]4CC[C@](O)(C(=O)CO)[C@@]4(C)C[C@H](O)[C@H]23

InChI

1S/C21H30O5/c1-19-7-5-13(23)9-12(19)3-4-14-15-6-8-21(26,17(25)11-22)20(15,2)10-16(24)18(14)19/h9,14-16,18,22,24,26H,3-8,10-11H2,1-2H3/t14-,15-,16-,18+,19-,20-,21-/m0/s1

InChI key

JYGXADMDTFJGBT-VWUMJDOOSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Hydrocortisone has been used in epithelial and endothelial adherent cell culture applications. Hydrocortisone solution has been used for the isolation of epidermal stem cells and cell expansion. It has also been used for keratinocyte permeability assay.

Other Notes

Animal-component free

Physical form

frozen in working aliquots, avoid repeated freeze/thaw

related product

Product No.
Description
Pricing

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A combination of aloe vera, curcumin, vitamin C, and taurine increases canine fibroblast migration and decreases tritiated water diffusion across canine keratinocytes in vitro.
Timothy R Fray et al.
The Journal of nutrition, 134(8 Suppl), 2117S-2119S (2004-07-31)
Natalja E Fedorovich et al.
Biomacromolecules, 10(7), 1689-1696 (2009-05-19)
Application of hydrogels in tissue engineering and innovative strategies such as organ printing, which is based on layered 3D deposition of cell-laden hydrogels, requires design of novel hydrogel matrices. Hydrogel demands for 3D printing include: 1) preservation of the printed
Xiao-hua Lei et al.
PloS one, 6(11), e26603-e26603 (2011-11-19)
The skin is susceptible to different injuries and diseases. One major obstacle in skin tissue engineering is how to develop functional three-dimensional (3D) substitute for damaged skin. Previous studies have proved a 3D dynamic simulated microgravity (SMG) culture system as
Shilpa Chennakrishnaiah et al.
Scientific reports, 10(1), 8532-8532 (2020-05-24)
Oncogenic transformation impacts cancer cell interactions with their stroma, including through formation of abnormal blood vessels. This influence is often attributed to angiogenic growth factors, either soluble, or associated with tumor cell-derived extracellular vesicles (EVs). Here we examine some of
Hongjuan Zhang et al.
Oncotarget, 7(3), 3367-3378 (2015-12-15)
Lung cancer is the global leading cause of cancer-related deaths. A significant portion of lung cancer patients harbor kinase domain mutations in the epidermal growth factor receptor (EGFR). While EGFR tyrosine kinase inhibitors (TKI) effectively shrink tumors harboring mutant EGFR

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service