SRP3292
R-SPONDIN-1 human
recombinant, expressed in CHO cells, ≥95% (SDS-PAGE), ≥95% (HPLC)
Synonym(s):
Rspo1
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352202
NACRES:
NA.32
Recommended Products
biological source
human
recombinant
expressed in CHO cells
Assay
≥95% (HPLC)
≥95% (SDS-PAGE)
form
lyophilized
potency
1.0-3.0 μg/mL ED50
mol wt
26.0-40.0 kDa
packaging
pkg of 20 μg
impurities
<0.1 EU/μg endotoxin, tested
color
white
UniProt accession no.
shipped in
wet ice
storage temp.
−20°C
Gene Information
human ... RSPO1(284654)
General description
R-Spondin-1 (Rspo-1) belongs to the (Rspo) family of Wnt modulators. Currently, the (Rspo) family consists of four structurally related secreted ligands (Rspo 1-4), all containing furin-like and thrombospondin structural domains. Rspo-1 is expressed in certain areas of the developing central nervous system, as well as in adrenal glands, ovary, testis, thyroid, and trachea. Recombinant human R-Spondin-1 is a 26.7kDa protein consisting of 243 amino acid residues. Due to glycosylation, R-Spondin-1 migrates at an apparent molecular weight of approximately 40.0kDa by SDS PAGE analysis under reducing conditions.
Biochem/physiol Actions
Rspo (R-Spondin) can interact with the Frizzled/LRP6 (LDL receptor related protein 6) receptor complex in a manner that stimulates the Wnt/beta-catenin signaling pathway. Disruption of Rspo-1 gene causes recessive syndrome with XX sex reversal, palmoplantar hyperkeratosis and predisposition to squamous cell carcinoma of the skin. Rspo-1 is also associated with radioresistance of tumor cells.
Physical form
Lyophilized from 10 mM Sodium Phosphate, pH 7.5.
Reconstitution
Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 0.1-1.0 mg/mL. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein (example 0.1% BSA) and store in working aliquots at -20°C to -80°C.
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
R-spondin1 is essential in sex determination, skin differentiation and malignancy.
Parma P, et al.
Nature Genetics, 38, 1304-1309 (2006)
R-Spondin proteins: a novel link to beta-catenin activation.
Kim KA, et al.
Cell Cycle, 5, 23-26 (2006)
Xuefeng Gu et al.
Oncotarget, 6(12), 9756-9765 (2015-04-14)
Although radiation therapy is the most effective postoperative adjuvant treatment, it does not substantially improve the long-term outcomes of glioma patients because of the characteristic radioresistance of glioma. We found that R-Spondin1 (Rspo1) expression was elevated in high-grade gliomas and
Minke E Binnerts et al.
Proceedings of the National Academy of Sciences of the United States of America, 104(37), 14700-14705 (2007-09-07)
The R-Spondin (RSpo) family of secreted proteins act as potent activators of the Wnt/beta-catenin signaling pathway. We have previously shown that RSpo proteins can induce proliferative effects on the gastrointestinal epithelium in mice. Here we provide a mechanism whereby RSpo1
Yong-Guo Zhang et al.
Autophagy, 15(11), 1935-1953 (2019-03-22)
Extracellular HMGB1 (high mobility group box 1) is considered as a damage-associated molecular pattern protein. However, little is known about its intracellular role. We studied the mechanism whereby intestinal epithelial HMGB1 contributes to host defense, using cell culture, colonoids, conditional
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service