Skip to Content
Merck
All Photos(2)

Documents

09-162

Sigma-Aldrich

Anti-Wnt3A Antibody

Upstate®, from rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human, mouse, rat

manufacturer/tradename

Upstate®

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... WNT3A(89780)

General description

The Wnt family of consists of ligands involved in cell development, fate determination, organogenesis, and oncogenesis. Information on Wnt signaling has been inferred from genetic analysis of wingless (wg) signaling in Drosophila. The Wnt signaling cascade initiates at the cell membrane when Wnt interacts with its receptor, a member of the Frizzled (Fz) transmembrane receptor family and lipoprotein receptor–related proteins 5 and 6 (LRP-5/6). This interaction relays the signal into the interior of the cell where disheveled (dsh) is brought to the membrane and gets phosphorylated/activated. Dsh promotes the inactivation of GSK3 kinase activity, thus preventing the phosphorylation of β-Catenin and allowing it to accumulate as it avoids ubiquination and subsequent proteosome degradation. Stable β-Catenin accumulates in the cytoplasm where it interacts with lymphoid enhancer factor/T cell factor (LEF/TCF) and translocates into the nucleus. This results in activation of TCF-responsive genes which are thought to play key roles in development and cancer. Negative regulators of the pathway may include axin and the tumor suppressor APC.

Specificity

Recognizes Wnt3A

Immunogen

Epitope: Central
Synthetic KLH-conjugated peptide directed against the central region of human, mouse, and rat Wnt3A.

Application

Anti-Wnt3A Antibody is an antibody against Wnt3A for use in WB, IH(P).
Research Category
Signaling
Research Sub Category
Developmental Signaling

Quality

routinely evaluated by immunoblot on rat brain lysate

Target description

~40 kDa

Physical form

Affinity purified
Affinity purified rabbit polyclonal in storage buffer containing PBS and 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.
Handling Recommendations: Upon receipt, and prior to removing the cap, centrifuge the vial and gently mix the solution.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kristina Rydell-Törmänen et al.
Physiological reports, 4(5) (2016-03-22)
Idiopathic pulmonary fibrosis (IPF), an insidious disease with grave prognosis, is characterized by heterogeneous fibrosis with densely fibrotic areas surrounded by nonfibrotic normal-looking tissue, believed to reflect a temporal development. The etiology is incompletely elucidated, but aberrant wound healing is
Ruoxi Liao et al.
Nephrology (Carlton, Vic.), 25(3), 264-272 (2019-11-12)
Vascular calcification (VC) is a common complication in chronic kidney disease (CKD) and has been shown to be associated with increased cardiovascular events and mortality. This study was to explore the role of Wnt-signaling pathway in CKD VC, and the
Xin Wang et al.
Neural regeneration research, 11(12), 2004-2011 (2017-02-16)
Electroacupuncture at Dazhui (GV14) and Mingmen (GV4) on the Governor Vessel has been shown to exhibit curative effects on spinal cord injury; however, the underlying mechanism remains poorly understood. In this study, we established rat models of spinal cord injury
Katarzyna Modzelewska et al.
Development (Cambridge, England), 147(14) (2020-06-27)
Long-term survival of an animal species depends on development being robust to environmental variations and climate changes. We used C. elegans to study how mechanisms that sense environmental changes trigger adaptive responses that ensure animals develop properly. In water, the
Guido Carpino et al.
PloS one, 11(6), e0157246-e0157246 (2016-06-17)
Non-alcoholic fatty liver disease is one of the most important causes of liver-related morbidity in children. In non-alcoholic fatty liver disease, the activation of liver resident macrophage pool is a central event in the progression of liver injury. The aims

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service