Skip to Content
Merck
All Photos(3)

Key Documents

SML1046

Sigma-Aldrich

CHIR99021

≥98% (HPLC), powder, GSK-3 inhibitor

Synonym(s):

6-[[2-[[4-(2,4-Dichlorophenyl)-5-(5-methyl-1H-imidazol-2-yl)-2-pyrimidinyl]amino]ethyl]amino]-3-pyridinecarbonitrile, CT99021

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H18Cl2N8
CAS Number:
Molecular Weight:
465.34
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Product Name

CHIR99021, ≥98% (HPLC)

Assay

≥98% (HPLC)

form

powder

color

white to light brown

solubility

DMSO: 2 mg/mL, clear (warmed)

storage temp.

−20°C

SMILES string

ClC(C=C1Cl)=CC=C1C2=NC(NCCNC(N=C3)=CC=C3C#N)=NC=C2C4=NC(C)=CN4

InChI

1S/C22H18Cl2N8/c1-13-10-29-21(31-13)17-12-30-22(32-20(17)16-4-3-15(23)8-18(16)24)27-7-6-26-19-5-2-14(9-25)11-28-19/h2-5,8,10-12H,6-7H2,1H3,(H,26,28)(H,29,31)(H,27,30,32)

InChI key

AQGNHMOJWBZFQQ-UHFFFAOYSA-N

General description

CHIR99021 is an aminopyrimidine derivative. CHIR99021 promotes self-renewal potential of embryonic stem cells (ESCs) of mice by inhibiting glycogen synthase kinase-3 (GSK-3) activity and potentiates the upregulation of β-catenin and c-Myc functions. CHIR99021 promotes self-renewal of ESCs by modulating transforming growth factor β (TGF-β), Notch and mitogen-activated protein kinases (MAPK) signaling pathways. CHIR99021 is an agonist of wingless/integrated (wnt) signalling, upregulates cyclinA expression and promotes cell proliferation in non-small-cell lung cancer (NSCLC) cell lines.

Application

CHIR99021 has been used as a Wnt signaling agonist to study its effects on the differentiation of human pluripotent stem cell-derived cardiomyocytes (hPSCs).
CHIR99021 has been used:
  • in cocktail with small molecules to convert human lung fibroblast cells into neurons
  • as a culture medium supplement in potassium supplemented simplex optimization medium (KSOM) for the culture of blastocysts
  • for chronic inhibition of GSK-3 to investigate its role in corticogenesis

Biochem/physiol Actions

CHIR99021 is a very potent inhibitor of Glycogen synthase kinase 3 (GSK3, IC50 values for GSK3α and GSK3β = 10 and 6.7 nM, respectively). There is no cross-reactivity against CDKs. CHIR99201 potentiates self-renewal of human and mouse embryonic stem cells, and enhances reprogramming of somatic cells into stem cells.
CHIR99021 is a very potent inhibitor of Glycogen synthase kinase 3.

also commonly purchased with this product

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The roles of Notch3 on the cell proliferation and apoptosis induced by CHIR99021 in NSCLC cell lines: a functional link between Wnt and Notch signaling pathways
Li C, et al.
PLoS ONE, 8(12), e84659-e84659 (2013)
CHIR99021 promotes self-renewal of mouse embryonic stem cells by modulation of protein-encoding gene and long intergenic non-coding RNA expression
Wu Y, et al.
Experimental Cell Research, 319(17), 2684-2699 (2013)
Co-incident insertion enables high efficiency genome engineering in mouse embryonic stem cells
Shy BR
Nucleic Acids Research, 44, 7997-8010 (2016)
Enterochromaffin Cells Are Gut Chemosensors that
Couple to Sensory Neural Pathways
Bellono NW
Cell, 170, 185-198 (2017)
Chemical conversion of human lung fibroblasts into neuronal cells
Wan XY, et al.
International Journal of Molecular Medicine, 41(3), 1463-1468 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service