Skip to Content
Merck
All Photos(5)

Key Documents

G9422

Sigma-Aldrich

β-Glycerophosphate disodium salt hydrate

BioUltra, suitable for cell culture, suitable for plant cell culture, ≥99% (titration)

Synonym(s):

BGP, Glycerol 2-phosphate disodium salt hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(HOCH2)2CHOP(O)(ONa)2 · xH2O
CAS Number:
Molecular Weight:
216.04 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

biological source

synthetic (chemical)

Quality Level

product line

BioUltra

Assay

≥99% (titration)

form

powder

technique(s)

cell culture | mammalian: suitable
cell culture | plant: suitable

impurities

≤0.1% Insoluble matter
≤0.5 mol % L-α-isomer

color

white

mp

102-104 °C (lit.)

solubility

water: soluble

anion traces

chloride (Cl-): ≤0.05%
sulfate (SO42-): ≤0.05%

cation traces

Al: ≤0.001%
Ca: ≤0.001%
Cu: ≤0.0005%
Fe: ≤0.001%
K: ≤0.005%
Mg: ≤0.0005%
Pb: ≤0.001%
Zn: ≤0.0005%

SMILES string

O.[Na+].[Na+].OCC(CO)OP([O-])([O-])=O

InChI

1S/C3H9O6P.2Na.H2O/c4-1-3(2-5)9-10(6,7)8;;;/h3-5H,1-2H2,(H2,6,7,8);;;1H2/q;2*+1;/p-2

InChI key

ROPZSVKNEIIIDE-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Glycerophosphoric acid disodium salt hydrate, also known as BGP, is a versatile compound with applications across various research domains, including cell biology, biochemical research, and metabolomics. Recognized as an endogenous metabolite and a serine-threonine phosphatase inhibitor, BGP is extensively utilized in chitosan hydrogel formulations for drug delivery research. It serves as a phosphate source for cell growth and recombinant protein production, offering advantages in media formulations by reducing precipitation at higher pH levels. Additionally, BGP finds applications in micro-arc oxidation electrolyte solutions, contributing to the creation of photocatalytic coatings.
In cell biology, β-Glycerophosphate disodium salt plays a pivotal role as a phosphate group donor in matrix mineralization studies and accelerates calcification in vascular smooth muscle cells. It also promotes bone matrix mineralization when delivered to osteoblasts, providing a crucial source of phosphate ions. In the realm of biochemical research, BGP functions as a classical serine-threonine phosphatase inhibitor in kinase reaction buffers, offering broad-spectrum inhibition. Its role as an organic phosphate donor is particularly valuable in culture media for mesenchymal stem cell differentiation to osteoblast-type cells. Moreover, BGP is utilized to buffer M17 media for Lactococcus culture in recombinant protein expression. The comprehensive utility of BGP spans multiple areas of research, making it an essential component in studies encompassing cell biology, biochemical research, and metabolomics.

Application

β-Glycerophosphate is a classical serine-threonine phosphatase inhibitor used in kinase reaction buffers. BGP is often used in combination with other phosphatase/protease inhibitors for broad spectrum inhibition. It functions as an organic phosphate donor and has been used in culture media for mesenchymal stem cell differentiation to osteoblast-type cells. BGP is also used to buffer M17 media for Lactococcus culture in recombinant protein expression.
Initiates differentiation of bone-marrow-derived stem cells embedded in a natural collagen/chitosan gel designed for tissue engineering.

Features and Benefits

  • Versatile and adaptable for a wide variety of laboratory and research applications
  • Suitable for Cell culture and Plant cell culture
  • Tested to confirm low levels of heavy metal contamination, ensuring suitability for various applications
  • BioUltra Grade for your Cell Biology and Biochemical research

Quality

Low inorganic phosphate content. Mixtures of α- and β-glycerophosphate have historically been used in microbiological media but high levels of inorganic (free) phosphate has been associated with the irregular growth of bacteria and unwanted precipitates.

Other Notes

For additional information on our range of Biochemicals, please complete this form.

comparable product

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Douglas et al.
The Journal of applied bacteriology, 56(2), 321-326 (1984-04-01)
An easily prepared medium, originally designed for the cultivation of lactic phages, has been found to have much wider application. Experience in its use over a ten year period with a range of physiologically diverse bacteria, for teaching and research
Lucas R Brun et al.
SpringerPlus, 3, 676-676 (2014-12-19)
The aim of this study was to evaluate the effect of strontium ranelate (SrR) on bone mineral density (BMD) and bone turnover markers after 1 year of treatment. Additionally, the effect of SrR in bisphosphonate-naïve patients (BP-naïve) compared to patients previously
Anna Di Vito et al.
Cell transplantation, 28(1), 129-139 (2018-10-30)
The periodontal ligament displays a reservoir of mesenchymal stem cells which can account for periodontal regeneration. Despite the numerous studies directed at the definition of optimal culture conditions for long-term expansion of periodontal ligament stem cells (PDLSCs), no consensus has
A Herrera et al.
Materials science & engineering. C, Materials for biological applications, 103, 109760-109760 (2019-07-28)
In the design of macroporous biomaterial scaffolds, attention is payed predominantly to the readily accessible macroscopic mechanical properties rather than to the mechanical properties experienced by the cells adhering to the material. However, the direct cell mechanical environment has been
Yulia Dzhashiashvili et al.
Neurobiology of disease, 127, 527-544 (2019-03-30)
Amyotrophic lateral sclerosis (ALS) is a progressive neurodegenerative disease, characterized by motor neuron death in the brain and spinal cord. Mutations in the Cu/Zn superoxide dismutase (SOD1) gene account for ~20% of all familial ALS forms, corresponding to 1%-2% of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service