Skip to Content
Merck
All Photos(3)

Key Documents

G7504

Sigma-Aldrich

Guanosine 3′,5′-cyclic monophosphate

≥98% (HPLC), powder, intracellular mediator

Synonym(s):

cGMP, cyclic GMP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H12N5O7P
CAS Number:
Molecular Weight:
345.21
EC Number:
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.77

Product Name

Guanosine 3′,5′-cyclic monophosphate, ≥98% (HPLC), powder

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white

solubility

0.2 M NaHCO3: 50 mg/mL

storage temp.

−20°C

SMILES string

NC1=NC(=O)c2ncn(C3OC4COP(O)(=O)OC4C3O)c2N1

InChI

1S/C10H12N5O7P/c11-10-13-7-4(8(17)14-10)12-2-15(7)9-5(16)6-3(21-9)1-20-23(18,19)22-6/h2-3,5-6,9,16H,1H2,(H,18,19)(H3,11,13,14,17)

InChI key

ZOOGRGPOEVQQDX-UHFFFAOYSA-N

Biochem/physiol Actions

An important second messenger, cGMP is a major intracellular mediator of extracellular signals such as nitric oxide and natriuretic peptides. Cyclic GMP interacts with three types of intracellular receptor proteins: cGMP-dependent protein kinases, cGMP-regulated channels, and cGMP-regulated cyclic nucleotide phosphodiesterases. A primary action of elevated cGMP levels in vivo is the stimulation of cGMP-dependent protein kinase (PKG).

Features and Benefits

This compound is a featured product for ADME Tox and Cyclic Nucleotide research. Discover more featured ADME Tox and Cyclic Nucleotide products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jeanette Erdmann et al.
Nature, 504(7480), 432-436 (2013-11-12)
Myocardial infarction, a leading cause of death in the Western world, usually occurs when the fibrous cap overlying an atherosclerotic plaque in a coronary artery ruptures. The resulting exposure of blood to the atherosclerotic material then triggers thrombus formation, which
Lasse H Hansen et al.
The Journal of biological chemistry, 294(34), 12567-12578 (2019-06-13)
Atrial natriuretic peptide (ANP) is a peptide hormone that in response to atrial stretch is secreted from atrial myocytes into the circulation, where it stimulates vasodilatation and natriuresis. ANP is an important biomarker of heart failure where low plasma concentrations
Juliane W Schott et al.
Molecular therapy. Methods & clinical development, 14, 134-147 (2019-07-25)
Ex vivo retroviral gene transfer into CD34+ hematopoietic stem and progenitor cells (HSPCs) has demonstrated remarkable clinical success in gene therapy for monogenic hematopoietic disorders. However, little attention has been paid to enhancement of culture and transduction conditions to achieve reliable
Ko-Hsin Chin et al.
Acta crystallographica. Section D, Biological crystallography, 69(Pt 3), 352-366 (2013-03-23)
The mammalian ER protein STING (stimulator of interferon genes; also known as MITA, ERIS, MPYS or TMEM173) is an adaptor protein that links the detection of cytosolic dsDNA to the activation of TANK-binding kinase 1 (TBK1) and its downstream transcription
Raquel Maeso-Díaz et al.
Aging and disease, 10(4), 684-698 (2019-08-24)
Advanced chronic liver disease (aCLD) represents a major public health concern. aCLD is more prevalent and severe in the elderly, carrying a higher risk of decompensation. We aimed at understanding how aging may impact on the pathophysiology of aCLD in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service