Skip to Content
Merck
All Photos(4)

Key Documents

A6885

Sigma-Aldrich

Adenosine 3′,5′-cyclic monophosphate sodium salt monohydrate

≥98.0% (HPLC), powder

Synonym(s):

3′,5′-Cyclic AMP sodium salt, cAMP-Na

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H11N5NaO6P · H2O
CAS Number:
Molecular Weight:
369.20
Beilstein:
54612
MDL number:
UNSPSC Code:
41106305
eCl@ss:
32160414
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98.0% (HPLC)

form

powder

color

white

solubility

H2O: 10 mg/mL

storage temp.

−20°C

SMILES string

[Na+].Nc1ncnc2n(cnc12)[C@@H]3O[C@@H]4COP([O-])(=O)O[C@H]4[C@H]3O

InChI

1S/C10H12N5O6P.Na/c11-8-5-9(13-2-12-8)15(3-14-5)10-6(16)7-4(20-10)1-19-22(17,18)21-7;/h2-4,6-7,10,16H,1H2,(H,17,18)(H2,11,12,13);/q;+1/p-1/t4-,6-,7-,10-;/m1./s1

InChI key

BXJBFCKTIWRKMQ-MCDZGGTQSA-M

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Adenosine-3′,5′-cyclic monophosphate (cAMP) is an activator of cyclic-AMP-dependent protein kinase A (PKA). The cAMP/PKA signaling pathway has been shown to inhibit cell proliferation, induce differentiation and lead to apoptosis. Exogneous cAMP plays a role in the spontaneous activity and contraction of the heart muscle.

Biochem/physiol Actions

Naturally-occurring activator of cyclic-AMP-dependent protein kinase (PKA).
Naturally-occurring activator of cyclic-AMP-dependent protein kinase (PKA). cAMP is an important second messenger that is linked in many systems to neurotransmitter- or hormone-induced receptor stimulation. The cAMP/PKA signaling pathway has been shown to inhibit cell proliferation, induce differentiation and lead to apoptosis.

Features and Benefits

This compound is a featured product for Cyclic Nucleotide research. Click here to discover more featured Cyclic Nucleotide products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound is featured on the Phosphodiesterases and PKA & PKG pages of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michaela Beese et al.
BMC cell biology, 11, 68-68 (2010-09-09)
Endothelial tight and adherens junctions control a variety of physiological processes like adhesion, paracellular transport of solutes or trafficking of activated leukocytes. Formation and maintenance of endothelial junctions largely depend on the microenvironment of the specific vascular bed and on
W Tuganowski et al.
Polish journal of pharmacology and pharmacy, 30(4), 451-454 (1978-07-01)
Isotonic solution of cAMP-Na was applied into spontaneously beating auricular fibres by a cut-end method. Cyclic AMP enhanced the spontaneous rate (mean 48%) and increased the contractile tension (mean 297%). These results provided direct evidence that cAMP is involved in
U Dunzendorfer et al.
European urology, 8(6), 363-366 (1982-01-01)
Patients with a history of idiopathic calcium oxalate stones, but without current stone formation, show distinctly higher levels of serum cAMP (cyclic adenosine 3',5'-monophosphate) after rapid injection of an extreme concentration of parathyroid extract (PTE). Extreme parathyroid hormone (PTH) levels
Logan A Collier et al.
mBio, 11(6) (2020-11-26)
The filamentous fungus Neurospora crassa decomposes lignocellulosic biomass to generate soluble sugars as carbon sources. In this study, we investigated a role for heterotrimeric G-protein signaling in cellulose degradation. Loss of the Gα subunit genes gna-1 and gna-3, the Gβ
Corinne Giusti et al.
Experimental cell research, 315(1), 26-38 (2008-10-28)
Dictyostelium atg1- mutant cells provide an experimentally and genetically favorable model to study necrotic cell death (NCD) with no interference from apoptosis or autophagy. In such cells subjected to starvation and cAMP, induction by the differentiation-inducing factor DIF or by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service