Skip to Content
Merck
All Photos(1)

Key Documents

G8377

Sigma-Aldrich

Guanosine 5′-monophosphate disodium salt hydrate

from yeast, ≥99%

Synonym(s):

5′-GMP-Na2, 5′-Guanylic acid disodium salt hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H12N5Na2O8P · xH2O
CAS Number:
Molecular Weight:
407.18 (anhydrous basis)
Beilstein:
3586801
EC Number:
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.51

biological source

yeast

Quality Level

Assay

≥99%

form

powder

solubility

H2O: soluble 50 mg/mL, clear, colorless to faintly yellow

storage temp.

room temp

SMILES string

O.[Na+].[Na+].NC1=Nc2c(ncn2[C@@H]3O[C@H](COP([O-])([O-])=O)[C@@H](O)[C@H]3O)C(=O)N1

InChI

1S/C10H14N5O8P.2Na.H2O/c11-10-13-7-4(8(18)14-10)12-2-15(7)9-6(17)5(16)3(23-9)1-22-24(19,20)21;;;/h2-3,5-6,9,16-17H,1H2,(H2,19,20,21)(H3,11,13,14,18);;;1H2/q;2*+1;/p-2/t3-,5-,6-,9-;;;/m1.../s1

InChI key

MEKWJTZTMVQYSU-CYCLDIHTSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Guanosine 5′-monophosphate is suitable for use in:
  • the measurement of density and viscosity of nucleotide and along with the furanose sugar from 0.0004 to 0.0014mol/kg solution at 288.15, 293.15 and 298.15K at atmospheric pressure
  • the detection and validation of a transcreener assay for detection of AMP- and GMP-producing enzymes
  • the preparation of nucleotide standard during the quantitative profiling of nucleotides using capillary IC-MS/MS

Biochem/physiol Actions

Guanosine 5′-monophosphate (GMP) is a ribonucleoside monophospate which upon phosphorylation to GTP becomes incorporated into ribonucleic acids (RNAs) by various RNA polymerase(s). Guanosine-5′-monophosphate (GMP) can be used to study modulation of glutamatergic neurotransmission.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Robbin R Vernooij et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 24(22), 5790-5803 (2018-01-10)
The photodecomposition mechanism of trans,trans,trans-[Pt(N3 )2 (OH)2 (py)2 ] (1, py=pyridine), an anticancer prodrug candidate, was probed using complementary Attenuated Total Reflection Fourier Transform Infrared (ATR-FTIR), transient electronic absorption, and UV/Vis spectroscopy. Data fitting using Principal Component Analysis (PCA) and
Marcelo Ganzella et al.
Purinergic signalling, 8(1), 49-58 (2011-09-02)
Overstimulation of the glutamatergic system (excitotoxicity) is involved in various acute and chronic brain diseases. Several studies support the hypothesis that guanosine-5'-monophosphate (GMP) can modulate glutamatergic neurotransmission. The aim of this study was to evaluate the effects of chronically administered
Roberto Farina Almeida et al.
Pharmacology, biochemistry, and behavior, 96(3), 306-311 (2010-06-23)
The glutamatergic system has received considerable attention over the last few years as potential target to develop anxiolytic drugs. Guanine based purines (GBPs) play an important neurmodulatory effect in the glutamatergic system. Several studies have shown the ability of the
Matt Staeben et al.
Assay and drug development technologies, 8(3), 344-355 (2010-02-18)
Screening of AMP- and GMP-producing enzymes such as phosphodiesterases (PDEs), ligases, and synthetases would be simplified by the ability to directly detect unmodified nucleoside monophosphates. To address this need, we developed polyclonal and monoclonal antibodies that recognize AMP and GMP
Simone Molz et al.
Neurochemical research, 30(1), 83-89 (2005-03-11)
Guanosine-5'-monophosphate (GMP) was evaluated as a neuroprotective agent against the damage induced by glutamate in rat hippocampal slices submitted to glucose deprivation. In slices maintained under physiological conditions, glutamate (0.01 to 10 mM), Kainate, alpha-amino-3-hydroxi-5-methylisoxazole-propionic acid (AMPA), N-methyl-D-aspartate (NMDA), 1S,3R-aminocyclopentane-1,3-dicarboxylic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service