Skip to Content
Merck
All Photos(4)

Documents

D6375

Sigma-Aldrich

2′-Deoxyadenosine 5′-monophosphate

Sigma Grade, 98-100%

Synonym(s):

2′-Deoxy-AMP, 2′-Deoxyadenylic acid, dAMP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H14N5O6P
CAS Number:
Molecular Weight:
331.22
EC Number:
MDL number:
UNSPSC Code:
41106305
eCl@ss:
32160414
PubChem Substance ID:
NACRES:
NA.51

biological source

synthetic (organic)

grade

Sigma Grade

Assay

98-100%

form

powder

solubility

1 N NH4OH: 50 mg/mL, clear, colorless

storage temp.

−20°C

SMILES string

Nc1ncnc2n(cnc12)[C@H]3C[C@H](O)[C@@H](COP(O)(O)=O)O3

InChI

1S/C10H14N5O6P/c11-9-8-10(13-3-12-9)15(4-14-8)7-1-5(16)6(21-7)2-20-22(17,18)19/h3-7,16H,1-2H2,(H2,11,12,13)(H2,17,18,19)/t5-,6+,7+/m0/s1

InChI key

KHWCHTKSEGGWEX-RRKCRQDMSA-N

General description

N2′-Deoxyadenosine 5′-monophosphate is a nucleotide that serves as a building block for DNA synthesis.

Application

2’-deoxyadenosine-5’-monophosphatehas been used to study proteins via UV resonance Raman spectroscopy.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kathryn W Woodburn et al.
Antibiotics (Basel, Switzerland), 9(1) (2020-01-17)
Acne vulgaris, caused by the Gram-positive bacterium Cutibacterium acnes, is a prevalent dermatologic condition with substantial cutaneous and psychological morbidity. Mild acne is treated with topical antibiotics with more severe inflammatory forms requiring the prolonged use of oral antibiotics, resulting
Toxic effects and DNA transformation
Savkovic S J
Journal of Separation Science, 6, 15-19 (2018)
Katsuhiko Sumitomo et al.
Scientific reports, 9(1), 13947-13947 (2019-09-29)
The use of DNA markers has revolutionized selection in crop breeding by linkage mapping and QTL analysis, but major problems still remain for polyploid species where marker-assisted selection lags behind the situation in diploids because of its high genome complexity.
Kathryn W Woodburn et al.
Journal of fungi (Basel, Switzerland), 6(3) (2020-09-26)
Cutaneous invasive fungal wound infections after life-threatening dismounted complex blast injury (DCBI) and natural disasters complicate clinical care. These wounds often require aggressive repeated surgical debridement, can result in amputations and hemipelvectomies and have a 38% mortality rate. Given the
Gabriela Petroselli et al.
Organic & biomolecular chemistry, 5(17), 2792-2799 (2007-08-19)
UV-A radiation (320-400 nm) induces damages to the DNA molecule and its components through photosensitized reactions. Pterins, heterocyclic compounds widespread in biological systems, participate in relevant biological processes and are able to act as photosensitizers. We have investigated the photosensitization

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service