Skip to Content
Merck
All Photos(4)

Key Documents

G7251

Sigma-Aldrich

Glycine anhydride

≥99% (TLC)

Synonym(s):

2,5-Diketopiperazine, 2,5-Piperazinedione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H6N2O2
CAS Number:
Molecular Weight:
114.10
Beilstein:
112112
EC Number:
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.26

Product Name

Glycine anhydride, cyclic dipeptide

Quality Level

Assay

≥99% (TLC)

form

powder

color

white

mp

>300 °C (lit.)

SMILES string

O=C1CNC(=O)CN1

InChI

1S/C4H6N2O2/c7-3-1-5-4(8)2-6-3/h1-2H2,(H,5,8)(H,6,7)

InChI key

BXRNXXXXHLBUKK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Oxidation of cyclic dipeptides by photoinduced H-atom abstraction. A laser flash FT EPR and optical spectroscopy study.: The study examines the oxidation processes of cyclic dipeptides, focusing on the role of glycine anhydride in photoinduced hydrogen atom abstraction reactions (Tarábek et al., 2007).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Prudel et al.
Die Nahrung, 29(4), 381-389 (1985-01-01)
A HPLC method for the determination of Usal (Aspartame hydrochloride, L-aspartyl-L-phenylalanine methyl ester hydrochloride) and its decomposition products was elaborated. Aspartic acid, phenylalanine, phenylalanine methyl ester, aspartyl-phenylalanine, phenylalanyl-aspartic acid, 5-benzyl-3,6-dioxo-2-piperazineacetic acid (DOP) and Usal were separated on Separon SI C-18.
J Haginaka et al.
The Journal of pharmacy and pharmacology, 38(3), 225-226 (1986-03-01)
A sensitive, high-performance liquid chromatographic method has been developed for the determination of piperazine-2,5-dione, a new metabolite of ampicillin, in human urine. Piperazine-2,5-dione was separated from human urine on a C18-column using phosphate buffer-methanol (pH 3.5) as eluent. Subsequently, the
R Shukla et al.
Peptides, 15(8), 1471-1474 (1994-01-01)
Central administration of exogenous cyclo(His-Pro) (CHP) is known to produce hypothermia in rodents. In the present study, we examined the role of endogenous CHP in cold-induced hypothermia in the desert rat, Mastomys natalensis. The results of these studies show that
Liyun Zhang et al.
Amino acids, 43(1), 279-287 (2011-09-16)
Cyclic dipeptides, due to their chemical properties and various bioactivities, are very attractive for medicinal chemistry. Fragmentations of three simple cyclic dipeptides including cyclo(Gly-Gly), cyclo(Ala-Ala) and cyclo(Gly-Val) in the gas-phase are determined with synchrotron vacuum ultraviolet (VUV) photoionization mass spectrometry
A K Szardenings et al.
Journal of medicinal chemistry, 41(13), 2194-2200 (1998-06-19)
The discovery of a novel series of heterocyclic matrix metalloproteinase (MMPs) inhibitors is described. Published crystal structures of peptidyl hydroxamates bound to MMPs were the basis for the rational design of diketopiperazine (DKP) inhibitors. Combinatorial libraries were prepared and evaluated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service