Skip to Content
Merck
All Photos(2)

Documents

219703

Sigma-Aldrich

Hexylamine

99%

Synonym(s):

1-Aminohexane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)5NH2
CAS Number:
Molecular Weight:
101.19
Beilstein:
1731298
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

liquid

expl. lim.

2.1-9.3 %

refractive index

n20/D 1.418 (lit.)

bp

131-132 °C (lit.)

mp

−23 °C (lit.)

density

0.766 g/mL at 25 °C (lit.)

SMILES string

CCCCCCN

InChI

1S/C6H15N/c1-2-3-4-5-6-7/h2-7H2,1H3

InChI key

BMVXCPBXGZKUPN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Hexylamine can be used:
  • As an initiator to synthesize defined polypeptides by primary amine-initiated N-carboxyanhydride ring opening polymerization reaction.
  • As a reactant to modify alkanethiol monolayers at polycrystalline gold surfaces via amide bond formation reaction.
  • To functionalize the surface of MWCNT, graphene oxide, and polyurethanes. These functionalized composites materials find applications in absorption, CO2 capture, and as barrier materials.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1A

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

80.6 °F - closed cup

Flash Point(C)

27 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Transformer oil based multi-walled carbon nanotube-hexylamine coolant with optimized electrical, thermal and rheological enhancements
Amiri A, et al.
Royal Society of Chemistry Advances, 5(130), 107222-107236 (2015)
Adsorption behaviour of n-hexylamine at the Hg/water interphase and its comparison with a molecular model accounting for local order.
Carla M, et al.
J. Electroanal. Chem. Interfac. Electrochem., 197(1), 123-141 (1986)
Jiong Zou et al.
Macromolecules, 46(10), 4223-4226 (2013-06-25)
A facile N
Amide and ester surface attachment reactions for alkanethiol monolayers at gold surfaces as studied by polarization modulation Fourier transform infrared spectroscopy.
Duevel RV and Corn RM.
Analytical Chemistry, 64(4), 337-342 (1992)
Mariko Miyachi et al.
Chemical communications (Cambridge, England), 46(15), 2557-2559 (2010-05-08)
Photosensing performance of a system composed of photosystem I (PSI), vitamin K(1) (VK(1))-like molecular wire, and gold nanoparticles (AuNPs) in an aqueous solution was increased considerably by the addition of double surfactants, hexylamine and dodecylbenzenesulfonate.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service