Skip to Content
Merck
All Photos(2)

Documents

220000

Sigma-Aldrich

Iminodiacetic acid

98%, for peptide synthesis

Synonym(s):

2,2′-Azanediyldiacetic acid, Diglycine, IMDA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HN(CH2COOH)2
CAS Number:
Molecular Weight:
133.10
Beilstein:
878499
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.22

product name

Iminodiacetic acid, 98%

Assay

98%

form

solid

mp

243 °C (dec.) (lit.)

application(s)

peptide synthesis

SMILES string

OC(=O)CNCC(O)=O

InChI

1S/C4H7NO4/c6-3(7)1-5-2-4(8)9/h5H,1-2H2,(H,6,7)(H,8,9)

InChI key

NBZBKCUXIYYUSX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Iminodiacetic acid is a versatile chelating agent widely used in metal extraction and waste water treatment.

Application

Iminodiacetic acid can be used:
  • In the preparation of iminodiacetic acid (IDA)-type adsorbent for protein purification.
  • As a catalyst along with iron (FeCl3) complexes for methyl methacrylate (MMA) polymerization.
  • In the preparation of chelating resins with glycidyl methacrylate for the selective removal and recovery of metal ions from industrial waste water.
  • As a reagent for the preparation of pinene-derived iminodiacetic acid (PIDA), which is in turn used as a ligand for the synthesis of diastereoselective oxiranyl C(sp3) boronates from the corresponding olefins.
  • In the surface functionalization of multi-walled carbon nanotubes (MWCNTs) which is further utilized as a sorbent for the separation of heavy metal ions.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Iminodiacetic acid modified kenaf fiber for waste water treatment
Razak MA, et al.
International Journal of Biological Macromolecules, 112, 754-760 (2018)
Synthesis of chelating resins with iminodiacetic acid and its wastewater treatment application
Wang C-C, et al.
Journal of Applied Polymer Science, 84(7), 1353-1362 (2002)
Chromatographic separation of proteins on metal immobilized iminodiacetic acid-bound molded monolithic rods of macroporous poly (glycidyl methacrylate-co-ethylene dimethacrylate)
Luo Q, et al.
Journal of Chromatography A, 926(2), 255-264 (2001)
Preparation of iminodiacetic acid functionalized multi-walled carbon nanotubes and its application as sorbent for separation and preconcentration of heavy metal ions
Wang J, et al.
Journal of Hazardous Materials, 186(2-3), 1985-1992 (2011)
Laia Reverté et al.
Environmental science & technology, 47(1), 471-478 (2012-12-12)
In this work, magnetic particles (MPs) are used as supports for the immobilization of biorecognition molecules for the detection of microcystins (MCs). In one approach, a recombinant protein phosphatase 1 (PP1) has been conjugated to MPs via coordination chemistry, and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service