Skip to Content
Merck
All Photos(3)

Documents

G1002

Sigma-Aldrich

Gly-Gly

≥99% (titration)

Synonym(s):

Diglycine, Glycyl-glycine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2CH2CONHCH2COOH
CAS Number:
Molecular Weight:
132.12
Beilstein:
1765223
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26

Assay

≥99% (titration)

form

powder

color

white

useful pH range

7.5-8.9

pKa (25 °C)

8.2

mp

262-264 °C

application(s)

diagnostic assay manufacturing

SMILES string

NCC(=O)NCC(O)=O

InChI

1S/C4H8N2O3/c5-1-3(7)6-2-4(8)9/h1-2,5H2,(H,6,7)(H,8,9)

InChI key

YMAWOPBAYDPSLA-UHFFFAOYSA-N

Gene Information

human ... SLC15A1(6564)

Looking for similar products? Visit Product Comparison Guide

Application

Gly-Gly has been used as a buffer for the measurement of glucose-6-phosphate dehydrogenase (G6PDH) activity and as a component of luciferase assay buffer.

Biochem/physiol Actions

Gly-Gly or diglycine is the simplest dipeptide. It may possess proteolytic properties. Gly-gly is hydrolyzed by glycylglycine dipeptidase.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Primary Epithelial Cell Models for Cystic Fibrosis Research
Scott H. Randell
Methods in Molecular Biology null
Open questions in origin of life: experimental studies on the origin of nucleic acids and proteins with specific and functional sequences by a chemical synthetic biology approach
Katarzyna A
Computational and structural biotechnology journal, 9, e201402004-e201402004 (2014)
THE GLYCYLGLYCINE DIPEPTIDASES OF SKELETAL
MUSCLE AND HUMAN UTERUS*
EMIL L. SMITH
The Journal of Biological Chemistry (1947)
Zheng Ma et al.
Antimicrobial agents and chemotherapy, 53(7), 2857-2864 (2009-04-22)
Nephrotoxicity is an important limitation to the clinical use of colistin against Pseudomonas aeruginosa and other gram-negative pathogens. Previous work reported net tubular reabsorption of colistin by the kidney in vivo, but there is no knowledge of its disposition within
Namrata D Udeshi et al.
Molecular & cellular proteomics : MCP, 12(3), 825-831 (2012-12-26)
Detection of endogenous ubiquitination sites by mass spectrometry has dramatically improved with the commercialization of anti-di-glycine remnant (K-ε-GG) antibodies. Here, we describe a number of improvements to the K-ε-GG enrichment workflow, including optimized antibody and peptide input requirements, antibody cross-linking

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service