Skip to Content
Merck
All Photos(2)

Documents

G6776

Sigma-Aldrich

Genistein

from Glycine max (soybean), ~98% (HPLC)

Synonym(s):

4′,5,7-Trihydroxyisoflavone, 5,7-Dihydroxy-3-(4-hydroxyphenyl)-4H-1-benzopyran-4-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H10O5
CAS Number:
Molecular Weight:
270.24
Beilstein:
263823
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

biological source

Glycine max (soybean)

Quality Level

Assay

~98% (HPLC)

form

powder

mp

>280 °C

solubility

DMSO: soluble
ethanol: soluble

storage temp.

−20°C

SMILES string

Oc1ccc(cc1)C2=COc3cc(O)cc(O)c3C2=O

InChI

1S/C15H10O5/c16-9-3-1-8(2-4-9)11-7-20-13-6-10(17)5-12(18)14(13)15(11)19/h1-7,16-18H

InChI key

TZBJGXHYKVUXJN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Genistein is a nutraceutical isoflavnoid from soybean and is a phytoestrogen. It is derived from genistin.

Application

Genistein has been used:
  • as an anticancer agent to test its cytotoxicity in cervix adenocarcinoma cells
  • as a polyphenol to test its effect on reactive oxygen species in mitochondria from sperm
  • to test its radical scavenging effect on the UV irradiated in dermal and keloid fibroblasts

Biochem/physiol Actions

Genistein has antioxidant functionality and is effective in quenching reactive oxygen species. It is beneficial to human health.
Inhibitor of tyrosine protein kinase; competitive inhibitor of ATP in other protein kinase reactions. Antiangiogenic agent, down-regulates the transcription of genes involved in controlling angiogenesis.

Features and Benefits

This compound is featured on the Met page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Jessica N Peoples et al.
Biochemical and biophysical research communications, 495(4), 2547-2552 (2017-12-31)
Mouse embryos that lack the ability to produce the adrenergic hormones, norepinephrine (NE) and epinephrine (EPI), due to disruption of the dopamine beta-hydroxylase (Dbh-/-) gene inevitably perish from heart failure during mid-gestation. Since adrenergic stimulation is well-known to enhance calcium
Kazuki Kusuyama et al.
The spine journal : official journal of the North American Spine Society, 18(6), 1062-1069 (2018-01-23)
Spinal cord injury (SCI) commonly results not only in motor paralysis but also in the emergence of neuropathic pain (NeuP), both of which can impair the quality of life for patients with SCI. In the clinical field, it is well
Lin Yang et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 25(3), 928-936 (2010-12-04)
The cardiac voltage-gated Ca(2+) channel, Ca(v)1.2, mediates excitation-contraction coupling in the heart. The molecular composition of the channel includes the pore-forming α1 subunit and auxiliary α2/δ-1 and β subunits. Ca(2+) channel γ subunits, of which there are 8 isoforms, consist
Haolong Li et al.
Oncotarget, 6(24), 20474-20484 (2015-05-27)
Although the new generation of androgen receptor (AR) antagonists like enzalutamide (ENZ) prolong survival of metastatic castration-resistant prostate cancer (CRPC), AR-driven tumors eventually recur indicating that additional therapies are required to fully block AR function. Since DNA topoisomerase II (Topo
Vivek Barve et al.
Journal of medicinal chemistry, 49(13), 3800-3808 (2006-06-23)
The synthesis and characterization of Schiff base derivatives of 3-formylchromone 3-6 (FPA-120 to FPA-123), the minimal biologically active structural motif of soy isoflavone, genistein, and their copper(II) complexes 7-10 (FPA-124 to FPA-127) are reported here. These copper complexes possess distorted

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service