Skip to Content
Merck
All Photos(1)

Key Documents

B9938

Sigma-Aldrich

Calycosin

≥98% (HPLC)

Synonym(s):

3′,7-Dihydroxy-4′-methoxy-isoflavone, 3′-Hydroxyformononetin, Cyclosin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H12O5
CAS Number:
Molecular Weight:
284.26
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.25

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to light yellow

solubility

methanol: 1 mg/mL, clear, colorless

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

2-8°C

SMILES string

O=C1C(C(C=C2)=CC(O)=C2OC)=COC3=CC(O)=CC=C13

InChI

1S/C16H12O5/c1-20-14-5-2-9(6-13(14)19)15-8-12(18)11-4-3-10(17)7-16(11)21-15/h2-8,17,19H,1H3

InChI key

QAGGICSUEVNSGH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Calycosin is a plant estrogen exhibiting both agonistic and antagonistic attitude towards estrogen. It is an isoflavone, occurring as the primary component of the Radix astragali extract.

Application

Calycosin has been used:
  • to study its effect on the repression of glioblastomas by having an inhibitory effect on the expression of tyrosine-protein kinase Met (c-Met)
  • to analyze its effect on the growth of nasopharyngeal carcinoma cells by modulating the long non-coding RNA, Ewing sarcoma-associated transcript 1(EWSAT1)
  • to evaluate its effect on the left ventricular ejection fraction in rat model with myocardial ischemia

Biochem/physiol Actions

Calycosin exhibits various therapeutic properties such as anti-inflammatory, antioxidant, antiviral, and neuroprotective effects. It is shown to exercise its anticarcinogenic effects against the cluster of breast cancer, osteosarcoma, hepatocellular carcinoma, and colorectal cancer. It is also being promoted as a probable treatment for myocardial infarction due to its ability to hasten the process of angiogenesis and increase the expression of vascular endothelial growth factor (VEGF). It has been found to show hyperpermeability induced by the conditions of low osmolarity. Finally, calycosin is being suggested to be used as a prodrug for the treatment of hypoxia-related vascular diseases.
Isoflavone and phytoestrogen isolated froma traditional Chinese medicinal herb. Calycosin induces angiogenesis in HUVEC and promotes proliferation of estrogen-receptor positive cells in vivo and in vitro. Also functions as a vasorelaxant on rat thoracic aorta.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Dongqing Zhang et al.
Oxidative medicine and cellular longevity, 2012, 209843-209843 (2012-08-01)
Flavonoids are found in most parts of plants and have been shown to have multiple biological activities such as anticancer, anti-inflammation, antibacteria, antivirus, and immune-stimulation. Existing data showed that the total flavonoids of Astragalus (TFA) can provide biological system with
Dongli Zhang et al.
Archives of biochemistry and biophysics, 691, 108478-108478 (2020-07-28)
Calycosin, a functional phytoestrogen isoflavone isolated from Radix astragali, has been shown to possess multiple pharmacological properties including anti-cancer activity. However, up to now, the anti-cancer effect and the related mechanism of calycosin on cervical cancer (CC) cells have not
Guang Hu et al.
Xenobiotica; the fate of foreign compounds in biological systems, 42(3), 294-303 (2011-10-04)
Although zebrafish has become a popular animal model for drug discovery and screening, drug metabolism in zebrafish remains largely unknown. In this study, we probed the metabolic capability of zebrafish larvae with calycosin, one of the major isoflavone constituents of
Gao Junqing et al.
Journal of traditional Chinese medicine = Chung i tsa chih ying wen pan, 35(2), 160-167 (2015-05-16)
To evaluated the effect of calycosin on left ventricular ejection fraction and angiogenesis. Adult male Sprague-Dawley rats were randomly assigned into calycosin-treated groups (0.5, 1, 2, and 4 mg/kg qd), a dimethyl sulfoxide (DMSO), or a sham-operated control group. The
Yi Fan et al.
European journal of pharmacology, 481(1), 33-40 (2003-11-26)
The purpose of the present study was to examine the effects of calycosin, an isoflavonoid isolated from Astragali Radix, on the impairment of barrier function induced by hypoxia in cultured human umbilical vein endothelial cells. Hypoxia induced an increase in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service