Skip to Content
Merck

P9783

Porcine Serum

USA origin, sterile-filtered, suitable for cell culture

Synonym(s):

pig sera, pig serum, porcine sera

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.71
UNSPSC Code:
12352207
Biological source:
Porcine
Origin:
USA origin
Sterility:
sterile-filtered
Shipped in:
dry ice
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

Porcine

Quality Segment

sterility

sterile-filtered

composition

hemoglobin, ≤50 mg/dL

origin

USA origin

technique(s)

cell culture | mammalian: suitable

impurities

≤20 EU/mL endotoxin

shipped in

dry ice

storage temp.

−20°C

General description

The porcine serum comprises serum amyloid A (SAA), C-reactive protein (CRP), haptoglobin (Hp), and pig major acute phase protein (pig-MAP).

Application

Porcine Serum has been used:
  • in the neutralization of lipofectamine coated adeno-associated viruses (AAVs)
  • for inducing liver fibrosis in rats
  • as a standard negative reference in enzyme-linked immunosorbent assay (ELISA)
  • as a component of trypticase soy broth with 0.6% yeast extract (TSBYE) for culturing A. actinomycetemcomitans strains

Biochem/physiol Actions

Pig serum acute phase proteins (APP) show elevation with age. It is used for the culture of porcine islet cells.

Preparation Note

Collected from adult animals.

Analysis Note

Endotoxin tested


Still not finding the right product?

Explore all of our products under Porcine Serum


Storage Class

10 - Combustible liquids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable

PPE (personal protective equipment)

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library