Skip to Content
Merck
All Photos(1)

Key Documents

G6148

Sigma-Aldrich

Glasgow Minimum Essential Medium

With L-glutamine, without tryptose phosphate broth and sodium bicarbonate, powder, suitable for cell culture

Synonym(s):

BHK medium, GMEM

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352207
NACRES:
NA.75

Quality Level

form

powder

technique(s)

cell culture | mammalian: suitable

components

glucose: 4.5 g/L
phenol red: 0.0213 g/L
L-glutamine: 0.292 g/L
NaHCO3: no

shipped in

ambient

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Glasgow Minimum Essential Medium is suitable for the study of genetic factors affecting cell competence.

Application

Glasgow Minimum Essential Medium has been used:
  • in toxin entry assays
  • for 3 H-palmitate labeling
  • to culture embryonic stem cells

Quantity

Formulated to contain 12.5 grams of powder per liter of medium.

Reconstitution

Supplement with 2.75 g/L sodium bicarbonate and Tryptose phosphate broth solution (T 8159) at 100 ml/L of medium.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tatsushi Miyazaki et al.
PloS one, 8(8), e71645-e71645 (2013-08-14)
Tcl1 is highly expressed in embryonic stem (ES) cells, but its expression rapidly decreases following differentiation. To assess Tcl1's roles in ES cells, we generated Tcl1-deficient and -overexpressing mouse ES cell lines. We found that Tcl1 was neither essential nor
Keiichi Ito et al.
Genes & development, 35(9-10), 729-748 (2021-04-24)
The MED1 subunit has been shown to mediate ligand-dependent binding of the Mediator coactivator complex to multiple nuclear receptors, including the adipogenic PPARγ, and to play an essential role in ectopic PPARγ-induced adipogenesis of mouse embryonic fibroblasts. However, the precise
Wander de Jesus Jeremias et al.
PloS one, 12(6), e0178829-e0178829 (2017-06-18)
Once inside a vertebrate host after infection, individual schistosomula of the parasite Schistosoma mansoni find a new and complex environment, which requires quick adjustments for survival, such as those that allow it to avoid the innate immune response of the
Naiara Clemente Tavares et al.
Frontiers in microbiology, 11, 963-963 (2020-07-01)
Schistosomiasis is a neglected tropical disease (NTD) caused by helminthes from the Schistosoma genus. This NTD can cause systemic symptoms induced by the deposition of parasite eggs in the host liver, promoting severe complications. Functional studies to increase knowledge about
Tatsushi Miyazaki et al.
PloS one, 8(8), e71645-e71645 (2013-08-14)
Tcl1 is highly expressed in embryonic stem (ES) cells, but its expression rapidly decreases following differentiation. To assess Tcl1's roles in ES cells, we generated Tcl1-deficient and -overexpressing mouse ES cell lines. We found that Tcl1 was neither essential nor

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service