Skip to Content
Merck

Skip To

A9502

Ala-Ala

Synonym(s):

L-Alanyl-L-alanine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Linear Formula:
CH3CH(NH2)CONHCH(CH3)CO2H
CAS Number:
Molecular Weight:
160.17
UNSPSC Code:
12352209
NACRES:
NA.26
PubChem Substance ID:
EC Number:
217-751-7
Beilstein/REAXYS Number:
1724813
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥98% (TLC)

form

powder

color

white

mp

280-285 °C (lit.)

SMILES string

C[C@H](N)C(=O)N[C@@H](C)C(O)=O

InChI

1S/C6H12N2O3/c1-3(7)5(9)8-4(2)6(10)11/h3-4H,7H2,1-2H3,(H,8,9)(H,10,11)/t3-,4-/m0/s1

InChI key

DEFJQIDDEAULHB-IMJSIDKUSA-N

Gene Information

Application

Ala-Ala has been used as a peptide sample for chromatographic analysis.

Biochem/physiol Actions

L-Alanyl-L-alanine is used as a model dipeptide in physicochemical studies of processes such as the effects of pH (protonation) on conformation.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
A9627A0912A0878
form

powder

form

powder

form

powder

form

powder

assay

≥98% (TLC)

assay

≥98% (TLC)

assay

≥99%

assay

≥99% (TLC)

mp

280-285 °C (lit.)

mp

-

mp

-

mp

-

color

white

color

white

color

white to off-white

color

white

Gene Information

human ... CA1(759), CA2(760), SLC15A1(6564)
rat ... Tpp2(81815)

Gene Information

-

Gene Information

-

Gene Information

-

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Jirí Sebek et al.
The journal of physical chemistry. A, 113(27), 7760-7768 (2009-06-17)
Fine effects of the hydration, charge, and conformational structural changes in L-alanyl-L-alanine (Ala-Ala) dipeptide were studied with the aid of Raman and Raman optical activity (ROA) spectra. The spectra were recorded experimentally and analyzed by means of density functional computations.
Determination of theoretical retention times for peptides analyzed by reversed-phase high-performance liquid chromatography.
Dziuba J, et al.
Acta Scientiarum Polonorum. Technologia Alimentaria, 10(2) (2011)
Ladislav Benda et al.
The journal of physical chemistry. B, 113(15), 5273-5281 (2009-03-24)
The dependence of the effective chemical shielding anisotropy (effective CSA, Deltasigma(eff)) on the phi and psi peptide backbone torsion angles was calculated in the l-alanyl-l-alanine (LALA) peptide using the DFT method. The effects of backbone conformation, molecular charge including the



Global Trade Item Number

SKUGTIN
A9502-100MG04061832883397
A9502-1G04061826752210

Questions

Reviews

No rating value

Active Filters