Skip to Content
Merck
All Photos(7)

Key Documents

B4639

Sigma-Aldrich

Biotin

powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%

Synonym(s):

D-Biotin, Bios II, Coenzyme R, Vitamin B7, Vitamin H

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H16N2O3S
CAS Number:
Molecular Weight:
244.31
Beilstein:
86838
EC Number:
MDL number:
UNSPSC Code:
12352201
eCl@ss:
34058011
PubChem Substance ID:
NACRES:
NA.21

product line

BioReagent

Quality Level

Assay

≥99%

form

powder

mol wt

Mw 244.31 g/mol

technique(s)

cell culture | insect: suitable
cell culture | mammalian: suitable
cell culture | plant: suitable

color

white to off-white

mp

231-233 °C (lit.)

solubility

H2O: 0.2 mg/mL (Solubility increases with addition of 1 N NaOH.)

storage temp.

2-8°C

SMILES string

[H][C@]12CS[C@@H](CCCCC(O)=O)[C@@]1([H])NC(=O)N2

InChI

1S/C10H16N2O3S/c13-8(14)4-2-1-3-7-9-6(5-16-7)11-10(15)12-9/h6-7,9H,1-5H2,(H,13,14)(H2,11,12,15)/t6-,7-,9-/m0/s1

InChI key

YBJHBAHKTGYVGT-ZKWXMUAHSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Biotin is also called as B complex vitamin. It is a bicyclic compound, which has a molecular weight of 244.31 Da. Biotin serves as an important cofactor for mammalian carboxylases. It is found within the cells in free and bound pools.

Application

Biotin has been used:
  • as a M199 media supplement in in vitro culture system
  • as a component of SATO serum free medium for the isolation of cortical oligodendrocyte precursors (OPCs)
  • in biochemical precursors culture for de novo fatty acid synthesis
  • as a component in aggregate culture media

Biochem/physiol Actions

Essential vitamin that is important for amino acid and energy metabolism, and fatty acid synthesis. It is a prosthetic group in four mammalian carboxylase families and facilitates the binding and transfer of carbon dioxide.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Encyclopedia of dietary supplements (2010)
Real-time in vivo green fluorescent protein imaging of a murine leishmaniasis model as a new tool for Leishmania vaccine and drug discovery
Mehta S R, et al.
Clinical and Vaccine Immunology : CVI, 15(12), 1764-1770 (2008)
Preparation of rat brain aggregate cultures for neuron and glia development studies
Koito H and Li J
Journal of Visualized Experiments, 31 (2009)
Convergent functional genomics of oligodendrocyte differentiation identifies multiple autoinhibitory signaling circuits
Gobert R P, et al.
Molecular and cellular biology, 29(6), 1538-1553 (2009)
Biochemical precursor effects on the fatty acid production in cell suspension cultures of Theobroma cacao L.
Parra O, et al.
Plant Physiology and Biochemistry, 111, 59-66 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service