Skip to Content
Merck
All Photos(1)

Key Documents

F8192

Sigma-Aldrich

Fetal Bovine Serum

Australia origin, Heat Inactivated, sterile-filtered, suitable for cell culture, suitable for hybridoma

Synonym(s):

HI FBS, HI sera, HI serum, FBS, FCS, sera, serum

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352207
NACRES:
NA.71

biological source

bovine fetus

Quality Level

sterility

sterile-filtered

product line

FBS Classic

composition

Bovine IgG, ≤1 mg/mL
Hemoglobin, ≤20 mg/dL

origin

Australia origin

technique(s)

cell culture | hybridoma: suitable
cell culture | mammalian: suitable

impurities

≤10 EU/mL endotoxin

shipped in

dry ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

FBS remains a popular media supplement because it provides a wide array of functions in cell culture. FBS delivers nutrients, growth and attachment factors and protects cells from oxidative damage and apoptosis by mechanisms that are difficult to reproduce in serum-free media (SFM) systems.

Application

FBS derived from clotted blood is the most widely used undefined supplement in eucaryotic, especially mammalian, cell culture. It is typically used with the classical media that were developed in the 1950s and 1960s. These include many popular media such as Minimum Essential Medium (MEM); Dulbecco′s Modified Eagles Media (DME, DMEM); Iscove′s Modified Eagles Medium (IMDM); M199 and Roswell Park Memorial Institute medium, RPMI-1640.

Preparation Note

Prepared by heating at 56°C for 30 minutes.

This product is prepared by heating at 56°C for 30 minutes. Heating serum to 56°C for 30 minutes inactivates the complement system for immunoassays. Heat activation also inactivates other undetermined inhibitors of cell growth in culture.

Analysis Note

  • Endotoxin and hemoglobin tested
  • Tested for the presence of bacteria, virus, and mycoplasma
  • Triple filtered with 0.1 micron membrane under aseptic conditions

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Jiamin Huang et al.
Inflammation research : official journal of the European Histamine Research Society ... [et al.], 69(3), 321-330 (2020-02-07)
To reveal the systematic response of neutrophils to sepsis and to study the hub lncRNAs in sepsis. Neutrophils taken from the femur and tibia of male C57 BL/6 mice were used in this study. And neutrophils were treated for 0 h
Leon M Larcher et al.
Molecules (Basel, Switzerland), 24(13) (2019-07-10)
MicroRNAs (miRNAs) are short non-coding RNAs that are involved in the regulation of gene expression. Previous reports showed an over-expression of miRNA-21 (miR-21) in various cancer cells, and its up-regulation is closely related to cancer initiation, proliferation and metastasis. In
Marti Cabanes-Creus et al.
Molecular therapy. Methods & clinical development, 12, 71-84 (2018-12-12)
Adeno-associated virus (AAV) vectors have become one of the most widely used gene transfer tools in human gene therapy. Considerable effort is currently being focused on AAV capsid engineering strategies with the aim of developing novel variants with enhanced tropism
Hyun Bong Park et al.
Nature microbiology, 5(11), 1319-1329 (2020-07-29)
Escherichia coli is an important model organism in microbiology and a prominent member of the human microbiota1. Environmental isolates readily colonize the gastrointestinal tract of humans and other animals, and they can serve diverse probiotic, commensal and pathogenic roles in
Deenaz Zaidi et al.
Journal of pediatric gastroenterology and nutrition, 71(5), 624-632 (2020-10-24)
Host-microbial relationship is disrupted in inflammatory bowel diseases (IBD). We hypothesized that altered gut luminal microenvironment can impact microbial virulence in IBD, leading to disruption of homeostasis and disease. We investigated the relationship between gut microenvironment and microbial virulence. Intestinal

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service