Skip to Content
Merck
All Photos(1)

Key Documents

F1016

Sigma-Aldrich

Fenoterol hydrobromide

Synonym(s):

2-(3,5-Dihydroxyphenyl)-2-hydroxy-2′-(4-hydroxyphenyl)-1′-methyldiethylamine hydrobromide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H21NO4 · HBr
CAS Number:
Molecular Weight:
384.26
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.77

Quality Level

originator

Boehringer Ingelheim

SMILES string

Br[H].CC(Cc1ccc(O)cc1)NCC(O)c2cc(O)cc(O)c2

InChI

1S/C17H21NO4.BrH/c1-11(6-12-2-4-14(19)5-3-12)18-10-17(22)13-7-15(20)9-16(21)8-13;/h2-5,7-9,11,17-22H,6,10H2,1H3;1H

InChI key

SGZRQMALQBXAIQ-UHFFFAOYSA-N

Gene Information

human ... ADRB2(154)

Looking for similar products? Visit Product Comparison Guide

General description

Fenoterol hydrobromide is a sympathomimetic drug. It is used to treat asthma, bronchitis and emphysema. Fenoterol administration might be linked to liver damage. It has anti-inflammatory effects on eosinophil function.

Application

Fenoterol hydrobromide has been used in chromatography-tandem mass spectrometry (LC&-MS-MS) for specific and sensitive detection of β-agonists (BAG).

Biochem/physiol Actions

β2-adrenoceptor agonist; bronchodilator.

Features and Benefits

This compound was developed by Boehringer Ingelheim. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dorota Latek et al.
Journal of molecular modeling, 17(9), 2353-2366 (2011-03-03)
Cannabinoid and adrenergic receptors belong to the class A (similar to rhodopsin) G protein coupled receptors. Docking of agonists and antagonists to CB(1) and CB(2) cannabinoid receptors revealed the importance of a centrally located rotamer toggle switch and its possible
Rajib K Paul et al.
The Journal of pharmacology and experimental therapeutics, 343(1), 157-166 (2012-07-11)
Inhibition of cell proliferation by fenoterol and fenoterol derivatives in 1321N1 astrocytoma cells is consistent with β(2)-adrenergic receptor (β(2)-AR) stimulation. However, the events that result in fenoterol-mediated control of cell proliferation in other cell types are not clear. Here, we
Oxford Dictionary of Medicines, Jan Hawthorn, 2003: Oxford Dictionary of Medicines (2003)
Bart G J Dekkers et al.
The Journal of pharmacology and experimental therapeutics, 342(3), 780-787 (2012-06-12)
Airway remodeling, including increased airway smooth muscle (ASM) mass and contractility, contributes to increased airway narrowing in asthma. Increased ASM mass may be caused by exposure to mitogens, including platelet-derived growth factor (PDGF) and collagen type I, which induce a
Julia J Rucklidge et al.
Human psychopharmacology, 27(5), 440-454 (2012-07-12)
To compare two micronutrient (vitamins and minerals) formulas (Berocca™ and CNE™) and assess their impact on emotions and stress related to the 6.3 earthquake on February 22(nd) 2011 in Christchurch, New Zealand. 91 adults experiencing heightened anxiety or stress 2-3 months

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service