Skip to Content
Merck
All Photos(3)

Key Documents

E7881

Sigma-Aldrich

Emodin

from Frangula bark, ≥90% (HPLC), casein Kinase 2 (CK2) inhibitor

Synonym(s):

1,3,8-Trihydroxy-6-methyl-9,10-anthracenedione, 1,3,8-Tri­hydroxy-6-methyl­anthra­quinone, 6-Methyl-1,3,8-tri­hydroxy­anthra­quinone, Archin, Emodol, Frangula-emodin, Frangulic acid, Rheum emodin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H10O5
CAS Number:
Molecular Weight:
270.24
Beilstein:
1888141
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77
Pricing and availability is not currently available.

Product Name

Emodin, from Frangula bark, ≥90% (HPLC)

biological source

Frangula bark

Quality Level

Assay

≥90% (HPLC)

solubility

DMSO: soluble
aqueous base: soluble
ethanol: soluble

storage temp.

2-8°C

SMILES string

Cc1cc(O)c2C(=O)c3c(O)cc(O)cc3C(=O)c2c1

InChI

1S/C15H10O5/c1-6-2-8-12(10(17)3-6)15(20)13-9(14(8)19)4-7(16)5-11(13)18/h2-5,16-18H,1H3

InChI key

RHMXXJGYXNZAPX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Emodin is an anthraquinone derivative, extracted from rhubarb (Rheum palmatum).[1] It has antibacterial,[1] anti-neoplastic, anti-inflammatory and anti-angiogenesis properties. Emodin exhibits cytotoxic effects, apoptosis and stimulates the expression of hypoxia-inducible factor 1α, glutathione S-transferase P and N-acetyltransferase.[2] It prevents the growth of prostate cancer cells.[1] Emodin is used to treat constipation and hemorrhoid. It displays immunosuppressive and anti-diabetic effects.[3]

Application

Emodin has been used:
  • to investigate its regulatory mechanisms on lipopolysaccharide (LPS)-induced inflammatory injury in myocarditis[4]
  • as a reference standard for the development and validation of high performance liquid chromatography (HPLC)-photodiode array (PDA) assay method[3]
  • to inhibit Tau aggregation[5]
  • for direct binding assay[6]

Biochem/physiol Actions

Inhibitor of NF-κB activation and adhesion molecule expression. Casein Kinase 2 (CK2) inhibitor.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Sheng-Zhang Lin et al.
PloS one, 7(8), e42146-e42146 (2012-08-10)
Emodin has been showed to induce apoptosis of pancreatic cancer cells and inhibit tumor growth in our previous studies. This study was designed to investigate whether emodin could inhibit the angiogenesis of pancreatic cancer tissues and its mechanism. In accordance
Anticancer potential of emodin
Hsu SC and Chung JG
BioMedicine, 2(3), 108-116 (2012)
Casein kinase 2 (CK2)-mediated phosphorylation of Hsp90beta as a novel mechanism of rifampin-induced MDR1 expression
Kim SW, et al.
The Journal of Biological Chemistry, 290(27), 17029-17040 (2015)
Development and Validation of HPLC-PDA Assay method of Frangula emodin
Duca D and Farrugia C
Mediterranean Journal of Chemistry, 5(2), 374-386 (2016)
Emodin and aloe-emodin suppress breast cancer cell proliferation through ERalpha inhibition
Huang PH, et al.
Evidence-Based Complementary and Alternative Medicine : ECAM, 2013 (2013)

Questions

  1. Is this a food grade supplement quality product?

    1 answer
    1. This product is intended for research use only and not suitable for human or veterinary use. Currently, a food grade product is not available.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service