Skip to Content
Merck
All Photos(1)

Key Documents

17797

Sigma-Aldrich

Physcion

≥98.0% (TLC)

Synonym(s):

1,8-Dihydroxy-3-methoxy-6-methylanthraquinone, 6-O-Methylemodin, 6-Methoxychrysophanic acid, Emodin-3-methyl ether, Parietin, Rheochrysidin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H12O5
CAS Number:
Molecular Weight:
284.26
Beilstein:
6770499
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.25

Assay

≥98.0% (TLC)

antibiotic activity spectrum

fungi

SMILES string

COc1cc(O)c2C(=O)c3c(O)cc(C)cc3C(=O)c2c1

InChI

1S/C16H12O5/c1-7-3-9-13(11(17)4-7)16(20)14-10(15(9)19)5-8(21-2)6-12(14)18/h3-6,17-18H,1-2H3

InChI key

FFWOKTFYGVYKIR-UHFFFAOYSA-N

Gene Information

human ... ELA2(1991)

Looking for similar products? Visit Product Comparison Guide

Application

Physcion can be used to study antibiotics, antifungals, bioactive small molecules and cell biology. Physcion has been used in a study to test the cytotoxicity of juglanthraquinone C on human cancer cells. Physcion has also been used in a study that investigated the metabolic regulatory effects of Rheum undulatum in a high-fat diet model.

Biochem/physiol Actions

Anthraquinone-derivative found in medicinal herbs.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Simeon F Kouam et al.
Journal of natural products, 69(2), 229-233 (2006-02-28)
One new prenylated 1,4-anthraquinone and three new prenylated anthranols, named kengaquinone (1) and kenganthranols A (2), B (3), and C (4), were isolated from a hexane extract of the stem bark of Harungana madagascariensis. Six known compounds including anthraquinones, anthrones
Yao Yao et al.
Apoptosis : an international journal on programmed cell death, 17(8), 832-841 (2012-04-10)
Juglanthraquinone C (1,5-dihydroxy-9,10-anthraquinone-3-carboxylic acid, JC), a naturally occurring anthraquinone isolated from the stem bark of Juglans mandshurica, shows strong cytotoxicity in various human cancer cells in vitro. Here, we first performed a structure-activity relationship study of six anthraquinone compounds (JC
Phil-Dong Moon et al.
Molecules (Basel, Switzerland), 24(8) (2019-04-18)
Physcion is well known for the treatment of carcinoma. However, the therapeutic effect of physcion on atopic dermatitis (AD) through the inhibition of thymic stromal lymphopoietin (TSLP) level remains largely unknown. In this study, we investigated the anti-AD effect of
Jie Tian et al.
Journal of separation science, 35(1), 145-152 (2011-12-07)
In this paper, two methods based on organic solvent dispersive liquid-liquid microextraction (OS-DLLME) and ionic liquid dispersive liquid-liquid microextraction (IL-DLLME) coupled with high-performance liquid chromatography have been critically compared for analyzing emodin and its metabolites (aloe-emodin, anthraquinone-2-carboxylic acid, rhein, danthron
Zhenli Liu et al.
Planta medica, 77(16), 1855-1860 (2011-06-16)
The dried roots of Polygonum multiflorum Thunb. (Heshouwu) and their processed products (Zhi-heshouwu) are widely used in traditional Chinese medicine, yet their therapeutic effects are different. Previous investigations focused mainly on the differences between Heshouwu and Zhi-heshouwu in the contents

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service