Skip to Content
Merck
All Photos(1)

Key Documents

203290

Sigma-Aldrich

Bisindolylmaleimide I

A highly selective, cell-permeable, and reversible protein kinase C (PKC) inhibitor (IC₅₀ = 10 nM) that is structurally similar to staurosporine.

Synonym(s):

Bisindolylmaleimide I, 2-[1-(3-Dimethylaminopropyl)-1H-indol-3-yl]-3-(1H-indol-3-yl)-maleimide, Gö 6850, GF 109203X

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H24N4O2
CAS Number:
Molecular Weight:
412.48
MDL number:
UNSPSC Code:
12352111
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

deep orange

solubility

DMSO: 10 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

N1C(=O)C(=C(C1=O)c4c5c([nH]c4)cccc5)c2c3c([n](c2)CCCN(C)C)cccc3

InChI

1S/C25H24N4O2/c1-28(2)12-7-13-29-15-19(17-9-4-6-11-21(17)29)23-22(24(30)27-25(23)31)18-14-26-20-10-5-3-8-16(18)20/h3-6,8-11,14-15,26H,7,12-13H2,1-2H3,(H,27,30,31)

InChI key

QMGUOJYZJKLOLH-UHFFFAOYSA-N

General description

A highly selective cell-permeable protein kinase C (PKC) inhibitor (IC50 = 10 nM) that is structurally similar to staurosporine. Acts as a competitive inhibitor for the ATP-binding site of PKC. Shows high selectivity for PKCα, βI, βII, γ, δ, and ε isozymes. May inhibit protein kinase A at a much higher concentration (IC50 = 2 µM). Also inhibits telomerase activity in quercetin, H-89, or herbimycin A treated NPC-076 cells.
A highly selective, cell-permeable, and reversible protein kinase C (PKC) inhibitor (IC50 = 10 nM) that is structurally similar to staurosporine. Acts as a competitive inhibitor for the ATP-binding site of PKC. Shows high selectivity for PKCα-, βI-, βII-, γ-, δ-, and ε- isozymes. Potently inhibits GSK-3 in primary adipocyte lysates (IC50 = 360 nM) and in GSK-3β immunoprecipitates (IC50 = 170 nM). May inhibit protein kinase A at a much higher concentration (IC50 = 2 µM). A 1 mg/ml solution of Bisindolylmaleimide I (Cat. No. 203293) in anhydrous DMSO is also available.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
PKC
Product competes with ATP.
Reversible: yes
Target IC50: 10 nM against protein kinase C (PKC); 360 nM, 170 nM against GSK-3 in primary adipocyte lysates and in GSK-3β immunoprecipitates, respectively

Warning

Toxicity: Standard Handling (A)

Preparation Note

Further dilute with aqueous buffers just prior to use.

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). DMSO stock solutions are stable for up to 4 months at -20°C.

Other Notes

Hers, I., et al. 1999. FEBS Lett.460, 433.
Ku, W.-C., et al. 1997. Biochem. Biophys. Res. Commun. 241, 730.
Gekeler, V., et al. 1996. Br. J. Cancer 74, 897.
Kiss, Z., et al. 1995. Biochim. Biophys. Acta 1265, 93.
Toullec, D., et al. 1991. J. Biol. Chem. 266, 15771.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Juliana C Corrêa-Velloso et al.
iScience, 24(10), 103139-103139 (2021-10-15)
Extracellular agonists linked to inositol-1,4,5-trisphosphate (IP3) formation elicit cytosolic Ca2+ oscillations in many cell types, but despite a common signaling pathway, distinct agonist-specific Ca2+ spike patterns are observed. Using qPCR, we show that rat hepatocytes express multiple purinergic P2Y and
Kirk D Haan et al.
STAR protocols, 4(4), 102744-102744 (2023-11-24)
Here, we present a protocol for live-cell immunocytochemistry to demonstrate reversible translocation of ion channels to the neuronal cell surface. We describe steps for cell preparation and isolation, experimental treatment, antibody binding prior to fixation, specific pipetting techniques, troubleshooting, and
Monique Rascher et al.
Biomolecules, 10(10) (2020-10-18)
Medicinal mushrooms of the genus Hericium are known to produce secondary metabolites with homeostatic properties for the central nervous system. We and others have recently demonstrated that among these metabolites cyathane diterpenoids and in particular erinacine C possess potent neurotrophin
Nicolas Chofflet et al.
Frontiers in molecular neuroscience, 17, 1371145-1371145 (2024-04-04)
The prevailing model behind synapse development and specificity is that a multitude of adhesion molecules engage in transsynaptic interactions to induce pre- and postsynaptic assembly. How these extracellular interactions translate into intracellular signal transduction for synaptic assembly remains unclear. Here
Katharina Koch et al.
Frontiers in toxicology, 4, 816370-816370 (2022-03-18)
There is a call for a paradigm shift in developmental neurotoxicity (DNT) evaluation, which demands the implementation of faster, more cost-efficient, and human-relevant test systems than current in vivo guideline studies. Under the umbrella of the Organisation for Economic Co-operation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service