Skip to Content
Merck
All Photos(3)

Key Documents

D9780

Sigma-Aldrich

Lambda DNA Hind III Digest

for DNA electrophoresis

Synonym(s):

Hind III digested bacteria phage Lambda DNA

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41141827
NACRES:
NA.25

grade

for molecular biology

form

liquid

usage

100 uses (0.1 mg size)
1000 uses (1 mg size)
20 uses (20 μg size)
500 uses (0.5 mg size)

suitability

suitable for electrophoresis (DNA)

shipped in

dry ice

storage temp.

−20°C

General description

Sigma′s Lambda DNA Hind III Digest contains 8 fragments from 125 – 23,130 bp. Ten ul of the ladder should be diluted in gel loading buffer and then loaded in a single lane on an agarose or polyacrylamide gel.

Application

Suitable for size determination of dsDNA by electrophoresis with either agarose or polyacrylamide gels.

Components

Sigma′s Lambda DNA Hind III Digest is provided in a solution of 10 mM Tris-HCl (pH 8.0), with 1.0 mM EDTA.

Other Notes

Contains 8 fragments, 125–23,130 bp
For optimal resolution, the recommended agarose gel concentration is 0.75%.

Preparation Note

Bacteriophage lambda DNA was digested with EcoR I and Hind III to completion, then purified by phenol extraction.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service