Skip to Content
Merck
All Photos(2)

Key Documents

C3934

Sigma-Aldrich

Carbonic Anhydrase from bovine erythrocytes

lyophilized powder, ≥2,000 W-A units/mg protein

Synonym(s):

Carbonate Dehydratase, Carbonate Hydrolyase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54
Pricing and availability is not currently available.

biological source

bovine erythrocytes

form

lyophilized powder

specific activity

≥2,000 W-A units/mg protein

mol wt

30 kDa

application(s)

diagnostic assay manufacturing

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

Carbonic anhydrase from bovine erythrocytes (BCA) has been used to study the effect of removal of enzyme-bound metal ion, Zn2+, on aggregation behavior of the enzyme. Removal of metal ion by a chelator such as EDTA enhances the propensity of the enzyme to adopt the molten-globule state. This state of the enzyme is found to bind to the chaperone-like α-crystallin and prevent aggregation.[1] The enzyme from Sigma has been immobilized to electrochemical transducers in order to obtain develop an analytical device for dissolved CO2 measurement.[2] It has also been used for the thermodynamic analysis of conformational changes in BCA.[3]

Biochem/physiol Actions

Carbonic anhydrase is a zinc metalloenzyme that has a molecular weight of approximately 30,000 Da. The enzyme catalyzes the hydration of carbon dioxide to carbonic acid. It is involved in vital physiological and pathological processes such as pH and CO2 homeostasis, transport of bicarbonate and CO2, biosynthetic reactions, bone resorption, calcification, and tumorigenicity. Therefore, this enzyme is an important target for inhibitors with clinical applications for various pathologies such as glaucoma, epilepsy and Parkinson′s disease.[4]

Unit Definition

One Wilbur-Anderson (W-A) unit will cause the pH of a 0.02 M Trizma buffer to drop from 8.3 to 6.3 per min at 0°C. (One W-A unit is essentially equivalent to one Roughton-Booth unit.)

inhibitor

Product No.
Description
Pricing

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mikhail Krasavin et al.
Journal of enzyme inhibition and medicinal chemistry, 35(1), 165-171 (2019-11-23)
Testing of an expanded, 800-compound set of analogues of the earlier described Strecker-type α-aminonitriles (selected from publicly available Enamine Ltd. Screening Collection) in thermal shift assay against bovine carbonic anhydrase (bCA) led to further validation of this new class of
Use of carbonic anhydrase in electrochemical biosensors for dissolved CO2
Cammaroto C, et al.
Sensors and Actuators B, Chemical, 48(1-3), 439-447 (1998)
Fraser J Moss et al.
The Journal of physiology, 598(24), 5821-5856 (2020-09-25)
According to the HCO 3 - metabolon hypothesis, direct association of cytosolic carbonic anhydrases (CAs) with the electrogenic Na/HCO3 cotransporter NBCe1-A speeds transport by regenerating/consuming HCO 3 - . The present work addresses published discrepancies as to whether
Matthew B Kerby et al.
Biotechnology progress, 22(5), 1416-1425 (2006-10-07)
This paper presents a microchip-based system for collecting kinetic time-based information on protein refolding and unfolding. Dynamic protein conformational change pathways were studied in microchannel flow using a microfluidic device. We present a protein-conserving approach for quantifying refolding by dynamically
K Rajaraman et al.
The Journal of biological chemistry, 271(44), 27595-27600 (1996-11-01)
alpha-Crystallin, a multimeric protein, exhibits chaperone-like activity in preventing aggregation of several proteins. We have studied the chaperone-like activity of alpha-crystallin toward heat-induced aggregation of bovine and human carbonic anhydrase. Human carbonic anhydrase aggregates at 60 degrees C, while bovine

Questions

1–5 of 5 Questions  
  1. Dear who is concern, I would like to know which isoform of carbonic anhydrase this is.

    1 answer
    1. This product is a mixture of the CA-I and CA-I isozymes. The ratio has not been determined.

      Helpful?

  2. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

  3. What is the molecular weight of Product C3934, Carbonate Dehydratase?

    1 answer
    1. Because there are a number of closely related isoforms, the molecular weight typically used is approximately 30 kDa.  This product is expected to be a mixture of isoforms.

      Helpful?

  4. What is the purity of Product C3934, Carbonate Dehydratase?

    1 answer
    1. Since C3934 is a mixture of isoforms, no purity claim is made.  The product is packaged and offered based on its enzymatic activity.

      Helpful?

  5. Can stock solutions of Product C3934, Carbonate Dehydratase, be made and stored?

    1 answer
    1. Carbonic anhydrase solutions can be prepared in water or buffer at concentrations up to 10 mg/mL.  It is common for there to be some particulates or haziness in the solution.  No data on solution stability has been collected by Sigma-Aldrich, so we recommend solutions be prepared fresh for use.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service