Skip to Content
Merck

Skip To

D6293

pUC18 DNA Hae III Digest

for DNA electrophoresis

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

NACRES:
NA.25
UNSPSC Code:
12352202
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

liquid

usage

200 uses (0.1 mg size), 40 uses (20 μg size)

suitability

suitable for electrophoresis (DNA)

storage temp.

−20°C

General description

Sigma′s pUC18 DNA Hae III Digest contains 11 fragments from 11 - 587 bp. About three ul of the ladder should be diluted in gel loading buffer and then loaded in a single lane on an agarose or polyacrylamide gel.

Application

Suitable for size determination of dsDNA by electrophoresis with either agarose or polyacrylamide gels.
pUC18 DNA Hae III Digest has been used as a DNA marker to compare the internal control developed for the competitive polymerase chain reaction (PCR) for the detection of Butyrivibrio fibrisolvens in the rumen.

Preparation Note

Plasmid pUC18 DNA was digested with Hae III to completion, then purified by phenol extraction.

Other Notes

Contains 11 fragments, 11–587 bp
For optimal resolution, the recommended agarose gel concentration is 2.5%.
Sigma′s pUC18 DNA Hae III Digest is provided in a solution of 10 mM Tris-HCl (pH 8.0), with 1.0 mM EDTA.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
D9780D3812D3937
description

for DNA electrophoresis

description

for DNA electrophoresis

description

ready-to-use marker for DNA electrophoresis

description

ready-to-use marker for DNA electrophoresis

form

liquid

form

liquid

form

(liquid)

form

liquid

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

usage

200 uses (0.1 mg size), 40 uses (20 μg size)

usage

1000 uses (1 mg size), 500 uses (0.5 mg size), 20 uses (20 μg size), 100 uses (0.1 mg size)

usage

sufficient for 100 loads

usage

100 uses

suitability

suitable for electrophoresis (DNA)

suitability

suitable for electrophoresis (DNA)

suitability

suitable for electrophoresis (DNA)

suitability

suitable for electrophoresis (DNA)


Storage Class

10 - Combustible liquids

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable

WGK

WGK 3



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

Reviews

No rating value

Active Filters